Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30783137-30783723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF3
Entrez Gene ID
51379
Gene Name
cytokine receptor like factor 3
Gene Aliases
CREME-9, CREME9, CRLM9, CYTOR4, FRWS, p48.2
UniGene
Hs.649372
Gene Chromosome Location
Chr.17: 30782684-30824760 on Build GRCh38
Transcripts
NM_015986.3 NR_073118.1 XM_011524879.1

SNP Detail

SNP ID Chromosome Location
rs895677104 Chr.17: 30783283-30783283 on Build GRCh38
rs911401361 Chr.17: 30783480-30783480 on Build GRCh38
rs9911658 Chr.17: 30783414-30783414 on Build GRCh38
rs187582109 Chr.17: 30783339-30783339 on Build GRCh38
rs148019594 Chr.17: 30783532-30783532 on Build GRCh38
rs905666038 Chr.17: 30783453-30783453 on Build GRCh38
rs1044318753 Chr.17: 30783286-30783286 on Build GRCh38
rs1048960599 Chr.17: 30783471-30783471 on Build GRCh38
rs536719100 Chr.17: 30783418-30783418 on Build GRCh38
rs184172132 Chr.17: 30783406-30783406 on Build GRCh38
rs890027089 Chr.17: 30783234-30783234 on Build GRCh38
rs995959223 Chr.17: 30783625-30783625 on Build GRCh38
rs1039072595 Chr.17: 30783250-30783250 on Build GRCh38
rs927165338 Chr.17: 30783600-30783601 on Build GRCh38
rs775130575 Chr.17: 30783684-30783684 on Build GRCh38
rs369689866 Chr.17: 30783604-30783607 on Build GRCh38
rs1044227893 Chr.17: 30783450-30783450 on Build GRCh38
rs9911885 Chr.17: 30783515-30783515 on Build GRCh38
rs907774668 Chr.17: 30783610-30783611 on Build GRCh38
rs959518741 Chr.17: 30783350-30783350 on Build GRCh38
rs150564195 Chr.17: 30783612-30783612 on Build GRCh38
rs570225312 Chr.17: 30783190-30783190 on Build GRCh38
rs1053200418 Chr.17: 30783282-30783282 on Build GRCh38
rs901520416 Chr.17: 30783266-30783266 on Build GRCh38
rs1024317779 Chr.17: 30783652-30783652 on Build GRCh38
rs894998150 Chr.17: 30783580-30783580 on Build GRCh38
rs1050168583 Chr.17: 30783627-30783627 on Build GRCh38
rs979046738 Chr.17: 30783488-30783488 on Build GRCh38
rs930571078 Chr.17: 30783461-30783461 on Build GRCh38
rs1004536108 Chr.17: 30783474-30783474 on Build GRCh38
rs911355033 Chr.17: 30783631-30783631 on Build GRCh38
rs866918072 Chr.17: 30783598-30783598 on Build GRCh38
rs9912283 Chr.17: 30783355-30783355 on Build GRCh38
rs762250576 Chr.17: 30783243-30783243 on Build GRCh38
rs544770203 Chr.17: 30783604-30783604 on Build GRCh38
rs191231997 Chr.17: 30783672-30783672 on Build GRCh38
rs753864745 Chr.17: 30783633-30783633 on Build GRCh38
rs925903208 Chr.17: 30783229-30783229 on Build GRCh38
rs9910234 Chr.17: 30783490-30783490 on Build GRCh38
rs537270446 Chr.17: 30783382-30783382 on Build GRCh38
rs1054397 Chr.17: 30783178-30783178 on Build GRCh38
rs547848421 Chr.17: 30783327-30783327 on Build GRCh38
rs1040041050 Chr.17: 30783619-30783619 on Build GRCh38
rs373790750 Chr.17: 30783638-30783638 on Build GRCh38
rs959349780 Chr.17: 30783162-30783162 on Build GRCh38
rs984655753 Chr.17: 30783472-30783472 on Build GRCh38
rs9890558 Chr.17: 30783337-30783337 on Build GRCh38
rs192030662 Chr.17: 30783445-30783445 on Build GRCh38
rs977351477 Chr.17: 30783636-30783636 on Build GRCh38
rs977449565 Chr.17: 30783177-30783177 on Build GRCh38
rs775893629 Chr.17: 30783518-30783543 on Build GRCh38
rs545451847 Chr.17: 30783487-30783487 on Build GRCh38
rs544639504 Chr.17: 30783486-30783486 on Build GRCh38
rs570146401 Chr.17: 30783655-30783655 on Build GRCh38
rs1010866097 Chr.17: 30783651-30783651 on Build GRCh38
rs759251409 Chr.17: 30783383-30783383 on Build GRCh38
rs1032563411 Chr.17: 30783557-30783557 on Build GRCh38
rs918703228 Chr.17: 30783226-30783226 on Build GRCh38
rs557869116 Chr.17: 30783654-30783655 on Build GRCh38
rs956263363 Chr.17: 30783510-30783510 on Build GRCh38
rs931450771 Chr.17: 30783230-30783230 on Build GRCh38
rs1048595953 Chr.17: 30783232-30783232 on Build GRCh38
rs954278106 Chr.17: 30783400-30783400 on Build GRCh38
rs1012518190 Chr.17: 30783589-30783589 on Build GRCh38
rs917326999 Chr.17: 30783443-30783443 on Build GRCh38
rs1042069150 Chr.17: 30783523-30783523 on Build GRCh38
rs141728465 Chr.17: 30783544-30783544 on Build GRCh38
rs1024879964 Chr.17: 30783596-30783596 on Build GRCh38
rs991192735 Chr.17: 30783170-30783170 on Build GRCh38
rs183910421 Chr.17: 30783540-30783540 on Build GRCh38
rs930477801 Chr.17: 30783601-30783604 on Build GRCh38
rs567256969 Chr.17: 30783420-30783420 on Build GRCh38
rs187619163 Chr.17: 30783613-30783613 on Build GRCh38
rs918667212 Chr.17: 30783417-30783417 on Build GRCh38
rs934748149 Chr.17: 30783253-30783253 on Build GRCh38
rs543606822 Chr.17: 30783223-30783223 on Build GRCh38
rs772284519 Chr.17: 30783637-30783639 on Build GRCh38
rs535353764 Chr.17: 30783594-30783594 on Build GRCh38
rs994690821 Chr.17: 30783297-30783297 on Build GRCh38
rs12006 Chr.17: 30783634-30783634 on Build GRCh38
rs529578687 Chr.17: 30783290-30783290 on Build GRCh38
rs552595460 Chr.17: 30783459-30783459 on Build GRCh38
rs976928943 Chr.17: 30783373-30783373 on Build GRCh38
rs1019141051 Chr.17: 30783479-30783479 on Build GRCh38
rs905801996 Chr.17: 30783288-30783288 on Build GRCh38
rs901455484 Chr.17: 30783512-30783512 on Build GRCh38
rs577555638 Chr.17: 30783484-30783484 on Build GRCh38
rs191921162 Chr.17: 30783398-30783398 on Build GRCh38
rs746356501 Chr.17: 30783456-30783457 on Build GRCh38
rs917926372 Chr.17: 30783172-30783172 on Build GRCh38
rs531943890 Chr.17: 30783215-30783215 on Build GRCh38
rs901470074 Chr.17: 30783621-30783624 on Build GRCh38
rs989324419 Chr.17: 30783432-30783432 on Build GRCh38
rs117699887 Chr.17: 30783285-30783285 on Build GRCh38
rs111654135 Chr.17: 30783270-30783270 on Build GRCh38
rs145737073 Chr.17: 30783353-30783353 on Build GRCh38
rs946306702 Chr.17: 30783648-30783648 on Build GRCh38
rs984233770 Chr.17: 30783628-30783628 on Build GRCh38
rs9890555 Chr.17: 30783333-30783333 on Build GRCh38

View Full Product Details