Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41882972-41883486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACLY
Entrez Gene ID
47
Gene Name
ATP citrate lyase
Gene Aliases
ACL, ATPCL, CLATP
UniGene
Hs.387567
Gene Chromosome Location
Chr.17: 41866916-41930542 on Build GRCh38
Transcripts
NM_001096.2 NM_001303274.1 NM_001303275.1 NM_198830.1 XM_005257395.1 XM_017024688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3188016 c.2240C>T p.T747M Chr.17: 41883147-41883147 on Build GRCh38
3517582 c.2186G>T p.R729L Chr.17: 41883201-41883201 on Build GRCh38
6592167 c.2235C>T p.I745I Chr.17: 41883152-41883152 on Build GRCh38
7281524 c.2259_2261delCTC p.S754delS Chr.17: 41883126-41883128 on Build GRCh38
5513308 c.2212A>G p.K738E Chr.17: 41883175-41883175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782644102 Chr.17: 41883146-41883146 on Build GRCh38
rs1023208631 Chr.17: 41883414-41883414 on Build GRCh38
rs1802730 Chr.17: 41883150-41883150 on Build GRCh38
rs782490187 Chr.17: 41883135-41883135 on Build GRCh38
rs782714978 Chr.17: 41883426-41883426 on Build GRCh38
rs782645249 Chr.17: 41883272-41883272 on Build GRCh38
rs913746956 Chr.17: 41883239-41883239 on Build GRCh38
rs1014343129 Chr.17: 41883388-41883388 on Build GRCh38
rs187181039 Chr.17: 41883263-41883263 on Build GRCh38
rs782263566 Chr.17: 41883147-41883147 on Build GRCh38
rs782079263 Chr.17: 41883083-41883083 on Build GRCh38
rs782588191 Chr.17: 41883122-41883122 on Build GRCh38
rs781846872 Chr.17: 41883101-41883101 on Build GRCh38
rs529835637 Chr.17: 41883011-41883011 on Build GRCh38
rs147061437 Chr.17: 41883096-41883096 on Build GRCh38
rs960170151 Chr.17: 41883069-41883069 on Build GRCh38
rs550214817 Chr.17: 41883286-41883286 on Build GRCh38
rs141894095 Chr.17: 41883316-41883316 on Build GRCh38
rs782425665 Chr.17: 41883152-41883152 on Build GRCh38
rs929421360 Chr.17: 41883354-41883354 on Build GRCh38
rs1038198074 Chr.17: 41882998-41882998 on Build GRCh38
rs782088357 Chr.17: 41883223-41883223 on Build GRCh38
rs782814548 Chr.17: 41883075-41883075 on Build GRCh38
rs782110532 Chr.17: 41883199-41883199 on Build GRCh38
rs782664431 Chr.17: 41883246-41883246 on Build GRCh38
rs782786524 Chr.17: 41883119-41883119 on Build GRCh38
rs534289739 Chr.17: 41883154-41883154 on Build GRCh38
rs191593638 Chr.17: 41883276-41883276 on Build GRCh38
rs1136265 Chr.17: 41883209-41883209 on Build GRCh38
rs376933182 Chr.17: 41883418-41883418 on Build GRCh38
rs1002299018 Chr.17: 41883440-41883440 on Build GRCh38
rs41275675 Chr.17: 41883182-41883182 on Build GRCh38
rs141297025 Chr.17: 41883201-41883201 on Build GRCh38
rs782371439 Chr.17: 41883270-41883270 on Build GRCh38
rs781910710 Chr.17: 41883247-41883247 on Build GRCh38
rs572699196 Chr.17: 41883298-41883298 on Build GRCh38
rs782522922 Chr.17: 41883245-41883245 on Build GRCh38
rs782527567 Chr.17: 41883107-41883107 on Build GRCh38
rs199569541 Chr.17: 41883226-41883226 on Build GRCh38
rs782224132 Chr.17: 41883269-41883269 on Build GRCh38
rs782433881 Chr.17: 41883279-41883279 on Build GRCh38
rs116646755 Chr.17: 41883250-41883250 on Build GRCh38
rs781927651 Chr.17: 41883077-41883077 on Build GRCh38
rs1026469937 Chr.17: 41883068-41883068 on Build GRCh38
rs782017025 Chr.17: 41883202-41883202 on Build GRCh38
rs782181799 Chr.17: 41883167-41883167 on Build GRCh38
rs782677992 Chr.17: 41883166-41883166 on Build GRCh38
rs201502123 Chr.17: 41883231-41883231 on Build GRCh38
rs1014478897 Chr.17: 41883091-41883091 on Build GRCh38
rs782219578 Chr.17: 41883131-41883131 on Build GRCh38
rs782750149 Chr.17: 41883225-41883225 on Build GRCh38
rs200343210 Chr.17: 41883121-41883121 on Build GRCh38
rs782160563 Chr.17: 41883208-41883208 on Build GRCh38
rs782337255 Chr.17: 41883181-41883181 on Build GRCh38
rs143028744 Chr.17: 41883033-41883033 on Build GRCh38
rs771760423 Chr.17: 41883415-41883415 on Build GRCh38
rs1048254215 Chr.17: 41883355-41883355 on Build GRCh38
rs79062040 Chr.17: 41883214-41883214 on Build GRCh38
rs1031991578 Chr.17: 41883451-41883451 on Build GRCh38
rs895992534 Chr.17: 41883386-41883386 on Build GRCh38

View Full Product Details