The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011512580.2 | XP_011510882.1 | 2 - 3 | 366 | 76 |
XM_011512581.2 | XP_011510883.1 | 2 - 3 | 366 | 76 | |
XM_011512584.2 | XP_011510886.1 | 2 - 3 | 366 | 76 | |
XM_017005939.1 | XP_016861428.1 | 2 - 3 | 396 | 76 | |
GenBank mRNA | BM504779.1 | - | 2 - 3 | 339 | 76 |