The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164595.1 | NP_001158067.1 | 2 - 3 | 738 | 112 |
GenBank mRNA | AK294535.1 | - | 2 - 3 | 201 | 112 |
AW297403.1 | - | 2 - 3 | 278 | 112 |