The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177533.4 | NP_803877.2 | 4 - 5 | 545 | 108 |
GenBank mRNA | AB087802.1 | - | 4 - 5 | 431 | 108 |
BC041584.1 | - | 4 - 5 | 507 | 108 |