Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASPDH
Entrez Gene ID
554235
Gene Name
aspartate dehydrogenase domain containing
Gene Aliases
-
Chromosome Location
Chr.19: 50511600 - 50514690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114598.1 NP_001108070.1 3 - 4 344 100
GenBank mRNA BM924033.1 - 3 - 4 344 100

View Full Product Details