The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198468.2 | NP_940870.2 | 21 - 22 | 3484 | 103 |
XM_006715430.1 | XP_006715493.1 | 21 - 22 | 3415 | 103 | |
XM_011535670.2 | XP_011533972.1 | 21 - 22 | 3415 | 103 | |
XM_011535671.2 | XP_011533973.1 | 21 - 22 | 3511 | 103 | |
XM_011535672.2 | XP_011533974.1 | 21 - 22 | 3478 | 103 | |
XM_011535673.2 | XP_011533975.1 | 21 - 22 | 3484 | 103 | |
XM_011535674.2 | XP_011533976.1 | 21 - 22 | 3380 | 103 | |
XM_011535675.2 | XP_011533977.1 | 20 - 21 | 3295 | 103 | |
XM_011535676.2 | XP_011533978.1 | 17 - 18 | 2879 | 103 | |
XM_011535677.2 | XP_011533979.1 | 14 - 15 | 2478 | 103 | |
XM_011535678.2 | XP_011533980.1 | 13 - 14 | 4100 | 103 | |
XM_011535679.2 | XP_011533981.1 | 11 - 12 | 2090 | 103 | |
XM_011535680.2 | XP_011533982.1 | 9 - 10 | 2254 | 103 | |
XM_017010656.1 | XP_016866145.1 | 20 - 21 | 3413 | 103 | |
XM_017010657.1 | XP_016866146.1 | 20 - 21 | 3402 | 103 | |
GenBank mRNA | AK128060.1 | - | 21 - 22 | 3484 | 103 |
CR749603.1 | - | 1 - 2 | 94 | 103 |