Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PROSER2
Entrez Gene ID
254427
Gene Name
proline and serine rich 2
Gene Aliases
C10orf47
Chromosome Location
Chr.10: 11823341 - 11872277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153256.3 NP_694988.3 1 - 2 73 75
GenBank mRNA BC017269.2 - 1 - 2 73 75
BC048800.1 - 1 - 2 73 75

View Full Product Details