Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRR32
Entrez Gene ID
100130613
Gene Name
proline rich 32
Gene Aliases
CXorf64
Chromosome Location
Chr.X: 126819764 - 126821786 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122716.1 NP_001116188.1 1 - 2 101 98
GenBank mRNA R10182.1 - - 101 98

View Full Product Details