The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080475.2 | NP_001073944.1 | 4 - 5 | 2189 | 73 |
XM_005246545.3 | XP_005246602.1 | 4 - 5 | 2189 | 73 | |
XM_006712521.3 | XP_006712584.1 | 4 - 5 | 2189 | 73 | |
XM_011511160.2 | XP_011509462.1 | 5 - 6 | 2524 | 73 | |
XM_011511163.2 | XP_011509465.1 | 4 - 5 | 2189 | 73 | |
XM_017004071.1 | XP_016859560.1 | 4 - 5 | 2189 | 73 | |
XM_017004072.1 | XP_016859561.1 | 3 - 4 | 2374 | 73 | |
XM_017004073.1 | XP_016859562.1 | 3 - 4 | 1253 | 73 | |
GenBank mRNA | AK123204.1 | - | 2 - 3 | 956 | 73 |
BC151137.1 | - | 3 - 4 | 1828 | 73 | |
BX648983.1 | - | 4 - 5 | 2128 | 73 |