Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41919694-41920276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf51
Entrez Gene ID
285636
Gene Name
chromosome 5 open reading frame 51
Gene Aliases
-
UniGene
Hs.29899
Gene Chromosome Location
Chr.5: 41904313-41921636 on Build GRCh38
Transcripts
NM_175921.5 XM_005248289.4 XM_011514032.2

SNP Detail

SNP ID Chromosome Location
rs140215964 Chr.5: 41920157-41920157 on Build GRCh38
rs147543931 Chr.5: 41920030-41920030 on Build GRCh38
rs991866718 Chr.5: 41919721-41919721 on Build GRCh38
rs965020972 Chr.5: 41920133-41920133 on Build GRCh38
rs548382767 Chr.5: 41919969-41919969 on Build GRCh38
rs987465319 Chr.5: 41920090-41920090 on Build GRCh38
rs1053983125 Chr.5: 41919833-41919833 on Build GRCh38
rs531289156 Chr.5: 41919824-41919824 on Build GRCh38
rs1026357389 Chr.5: 41920056-41920056 on Build GRCh38
rs952668177 Chr.5: 41920230-41920230 on Build GRCh38
rs1054787836 Chr.5: 41919958-41919958 on Build GRCh38
rs945531197 Chr.5: 41920200-41920200 on Build GRCh38
rs1049095509 Chr.5: 41919830-41919830 on Build GRCh38
rs748668982 Chr.5: 41920125-41920125 on Build GRCh38
rs931219969 Chr.5: 41919828-41919828 on Build GRCh38
rs1005863482 Chr.5: 41920006-41920006 on Build GRCh38
rs919747088 Chr.5: 41920196-41920196 on Build GRCh38
rs990154283 Chr.5: 41920153-41920153 on Build GRCh38
rs749586495 Chr.5: 41920027-41920027 on Build GRCh38
rs933730821 Chr.5: 41920158-41920158 on Build GRCh38
rs1003219238 Chr.5: 41919934-41919934 on Build GRCh38
rs1042179950 Chr.5: 41919872-41919872 on Build GRCh38
rs10051536 Chr.5: 41919841-41919841 on Build GRCh38
rs978221832 Chr.5: 41920227-41920227 on Build GRCh38
rs562934111 Chr.5: 41919842-41919842 on Build GRCh38
rs987391835 Chr.5: 41919725-41919725 on Build GRCh38
rs980638985 Chr.5: 41920179-41920179 on Build GRCh38
rs902034833 Chr.5: 41919912-41919912 on Build GRCh38
rs371324328 Chr.5: 41920226-41920226 on Build GRCh38
rs534243102 Chr.5: 41920147-41920147 on Build GRCh38
rs370446715 Chr.5: 41919819-41919819 on Build GRCh38
rs906244080 Chr.5: 41919876-41919876 on Build GRCh38
rs769040894 Chr.5: 41920085-41920085 on Build GRCh38
rs186465491 Chr.5: 41919763-41919763 on Build GRCh38
rs779344250 Chr.5: 41920119-41920119 on Build GRCh38
rs533735899 Chr.5: 41920036-41920036 on Build GRCh38
rs954726699 Chr.5: 41919724-41919724 on Build GRCh38
rs143298436 Chr.5: 41919966-41919966 on Build GRCh38
rs904147125 Chr.5: 41920029-41920030 on Build GRCh38

View Full Product Details