Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH2
Entrez Gene ID
121642
Gene Name
alkB homolog 2, alpha-ketoglutarate dependent dioxygenase
Gene Aliases
ABH2
Chromosome Location
Chr.12: 109088188 - 109093631 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001655.2 NP_001001655.1 2 - 3 524 86
NM_001145374.1 NP_001138846.1 2 - 3 674 86
NM_001145375.1 NP_001138847.1 2 - 3 421 86
XM_005253835.4 XP_005253892.1 2 - 3 690 86
GenBank mRNA AY754389.1 - 2 - 3 509 86
BC070489.1 - 2 - 3 476 86
BM790974.1 - 2 - 3 403 86

View Full Product Details