The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142964.1 | NP_001136436.1 | 1 - 2 | 299 | 54 |
XM_011530373.2 | XP_011528675.1 | 1 - 2 | 293 | 54 | |
XM_017028939.1 | XP_016884428.1 | 2 - 3 | 424 | 54 | |
GenBank mRNA | AK131457.1 | - | 1 - 2 | 275 | 54 |
BC035403.1 | - | 1 - 2 | 299 | 54 |