The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138471.2 | NP_612480.1 | 4 - 5 | 1049 | 73 |
XM_005273782.2 | XP_005273839.1 | 4 - 5 | 1049 | 73 | |
XM_005273783.2 | XP_005273840.1 | 4 - 5 | 1049 | 73 | |
XM_006718437.1 | XP_006718500.1 | 4 - 5 | 1049 | 73 | |
XM_011544770.1 | XP_011543072.1 | 4 - 5 | 1049 | 73 | |
XM_011544772.1 | XP_011543074.1 | 4 - 5 | 1049 | 73 | |
GenBank mRNA | BC002782.2 | - | 4 - 5 | 807 | 73 |
BC007540.1 | - | 4 - 5 | 807 | 73 | |
BC056402.1 | - | 4 - 5 | 972 | 73 |