Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11orf84
Entrez Gene ID
144097
Gene Name
chromosome 11 open reading frame 84
Gene Aliases
-
Chromosome Location
Chr.11: 63813374 - 63827718 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138471.2 NP_612480.1 5 - 6 1249 100
GenBank mRNA BC002782.2 - 5 - 6 1007 100
BC007540.1 - 5 - 6 1007 100
BC056402.1 - 5 - 6 1172 100
CR749697.1 - 1 - 2 68 100

View Full Product Details