Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17orf67
Entrez Gene ID
339210
Gene Name
chromosome 17 open reading frame 67
Gene Aliases
-
Chromosome Location
Chr.17: 56791913 - 56833990 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085430.2 NP_001078899.2 7 - 8 96
GenBank mRNA BC041467.1 - 7 - 8 96
CA308560.1 - 1 - 2 456 96

View Full Product Details