Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C5orf42
Entrez Gene ID
65250
Gene Name
chromosome 5 open reading frame 42
Gene Aliases
Hug, JBTS17, OFD6
Chromosome Location
Chr.5: 37063928 - 37249428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023073.3 NP_075561.3 11 - 12 1748 97
XM_005248345.3 XP_005248402.1 11 - 12 1736 97
XM_005248346.3 XP_005248403.1 11 - 12 1656 97
XM_005248347.3 XP_005248404.1 11 - 12 1656 97
XM_005248349.3 XP_005248406.1 11 - 12 1656 97
XM_005248350.3 XP_005248407.1 11 - 12 1654 97
XM_011514085.2 XP_011512387.1 11 - 12 1655 97
XM_011514086.2 XP_011512388.1 11 - 12 1652 97
XM_011514087.2 XP_011512389.1 11 - 12 1741 97
XM_011514088.2 XP_011512390.1 11 - 12 1741 97
XM_011514089.2 XP_011512391.1 11 - 12 1741 97
XM_011514090.2 XP_011512392.1 11 - 12 1519 97
XM_011514092.2 XP_011512394.1 11 - 12 1741 97
XM_017009760.1 XP_016865249.1 10 - 11 1467 97
XM_017009761.1 XP_016865250.1 10 - 11 1435 97
XM_017009762.1 XP_016865251.1 7 - 8 1194 97
XM_017009763.1 XP_016865252.1 7 - 8 1067 97
XM_017009764.1 XP_016865253.1 7 - 8 1028 97
XM_017009765.1 XP_016865254.1 11 - 12 1436 97
GenBank mRNA AW996738.1 - 1 - 2 97
DN831769.1 - 1 - 2 94 97

View Full Product Details