Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMY2A
Entrez Gene ID
279
Gene Name
amylase, alpha 2A (pancreatic)
Gene Aliases
AMY2, PA
Chromosome Location
Chr.1: 103617332 - 103625780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000699.3 NP_000690.1 1 - 2 281 120
GenBank mRNA AK223463.1 - 1 - 2 196 120
BC007060.1 - 1 - 2 236 120
BC146997.1 - 1 - 2 281 120
M28443.1 - 1 - 2 185 120

Gene Symbol
AMY1A
Entrez Gene ID
276
Gene Name
amylase, alpha 1A (salivary)
Gene Aliases
AMY1
Chromosome Location
Chr.1: 103655519 - 103664551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008221.1 NP_001008222.1 2 - 3 386 120
NM_004038.3 NP_004029.2 2 - 3 467 120
GenBank mRNA AK292341.1 - 2 - 3 385 120
AK313275.1 - 2 - 3 387 120
BC063129.1 - 2 - 3 396 120
BC069347.1 - 1 - 2 172 120
BC069463.1 - 1 - 2 172 120
BC070302.1 - 2 - 3 392 120
BC092444.1 - 2 - 3 363 120
BC132985.1 - 2 - 3 342 120
BC132987.1 - 2 - 3 342 120
BC132995.1 - 2 - 3 341 120
BC132997.1 - 2 - 3 342 120
BC144452.1 - 2 - 3 342 120
BX487266.1 - 2 - 3 387 120
CB956272.1 - 2 - 3 470 120

Gene Symbol
AMY1B
Entrez Gene ID
277
Gene Name
amylase, alpha 1B (salivary)
Gene Aliases
AMY1
Chromosome Location
Chr.1: 103687415 - 103696452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008218.1 NP_001008219.1 2 - 3 444 120
XM_011541262.1 XP_011539564.1 2 - 3 387 120

Gene Symbol
AMY1C
Entrez Gene ID
278
Gene Name
amylase, alpha 1C (salivary)
Gene Aliases
AMY1
Chromosome Location
Chr.1: 103745430 - 103758692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008219.1 NP_001008220.1 2 - 3 466 120
XM_011541263.1 XP_011539565.1 2 - 3 387 120
XM_017001058.1 XP_016856547.1 2 - 3 304 120
GenBank mRNA CB955582.1 - 2 - 3 254 120
CB985420.1 - 2 - 3 361 120
CB987307.1 - 2 - 3 469 120
CD690933.1 - 3 - 4 397 120

Gene Symbol
AMY2B
Entrez Gene ID
280
Gene Name
amylase, alpha 2B (pancreatic)
Gene Aliases
AMY2, AMY3, HXA
Chromosome Location
Chr.1: 103554644 - 103579534 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020978.4 NP_066188.1 3 - 4 844 120
GenBank mRNA AK095605.1 - 2 - 3 1514 120
AK127047.1 - 2 - 3 2283 120
BC011179.1 - 3 - 4 536 120
BC020861.1 - 2 - 3 308 120
M24895.1 - 1 - 2 218 120

View Full Product Details