Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6645086-6645540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
942785 c.388C>T p.L130F Chr.12: 6645307-6645307 on Build GRCh38
1704987 c.409G>A p.E137K Chr.12: 6645286-6645286 on Build GRCh38
4790176 c.365T>G p.L122R Chr.12: 6645330-6645330 on Build GRCh38
4829757 c.418G>C p.A140P Chr.12: 6645277-6645277 on Build GRCh38
4549553 c.475G>A p.V159M Chr.12: 6645220-6645220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558238595 Chr.12: 6645159-6645159 on Build GRCh38
rs761796073 Chr.12: 6645280-6645280 on Build GRCh38
rs10849493 Chr.12: 6645352-6645352 on Build GRCh38
rs984064832 Chr.12: 6645281-6645281 on Build GRCh38
rs565120830 Chr.12: 6645456-6645456 on Build GRCh38
rs765011108 Chr.12: 6645286-6645286 on Build GRCh38
rs950856003 Chr.12: 6645397-6645397 on Build GRCh38
rs371966000 Chr.12: 6645177-6645177 on Build GRCh38
rs765971089 Chr.12: 6645310-6645310 on Build GRCh38
rs374469627 Chr.12: 6645367-6645367 on Build GRCh38
rs771438305 Chr.12: 6645212-6645212 on Build GRCh38
rs932267196 Chr.12: 6645141-6645141 on Build GRCh38
rs577802329 Chr.12: 6645111-6645111 on Build GRCh38
rs141212919 Chr.12: 6645150-6645161 on Build GRCh38
rs774916783 Chr.12: 6645224-6645224 on Build GRCh38
rs181598197 Chr.12: 6645108-6645108 on Build GRCh38
rs751186544 Chr.12: 6645377-6645377 on Build GRCh38
rs181616785 Chr.12: 6645516-6645516 on Build GRCh38
rs764809840 Chr.12: 6645179-6645179 on Build GRCh38
rs550896657 Chr.12: 6645494-6645494 on Build GRCh38
rs747050351 Chr.12: 6645357-6645357 on Build GRCh38
rs754452777 Chr.12: 6645321-6645321 on Build GRCh38
rs35020360 Chr.12: 6645329-6645329 on Build GRCh38
rs762768769 Chr.12: 6645373-6645373 on Build GRCh38
rs758113346 Chr.12: 6645187-6645187 on Build GRCh38
rs368620092 Chr.12: 6645301-6645301 on Build GRCh38
rs959386028 Chr.12: 6645242-6645242 on Build GRCh38
rs749802705 Chr.12: 6645325-6645325 on Build GRCh38
rs973472422 Chr.12: 6645133-6645133 on Build GRCh38
rs758241196 Chr.12: 6645366-6645366 on Build GRCh38
rs750357050 Chr.12: 6645274-6645274 on Build GRCh38
rs11064298 Chr.12: 6645147-6645147 on Build GRCh38
rs750062153 Chr.12: 6645181-6645181 on Build GRCh38
rs12582432 Chr.12: 6645168-6645168 on Build GRCh38
rs758023830 Chr.12: 6645308-6645308 on Build GRCh38
rs778287779 Chr.12: 6645324-6645324 on Build GRCh38
rs772890592 Chr.12: 6645334-6645335 on Build GRCh38
rs200286554 Chr.12: 6645261-6645261 on Build GRCh38
rs772943133 Chr.12: 6645368-6645368 on Build GRCh38
rs188527378 Chr.12: 6645197-6645197 on Build GRCh38
rs775227814 Chr.12: 6645342-6645342 on Build GRCh38
rs748794301 Chr.12: 6645117-6645117 on Build GRCh38
rs367996082 Chr.12: 6645252-6645252 on Build GRCh38
rs765991933 Chr.12: 6645374-6645374 on Build GRCh38
rs548191198 Chr.12: 6645158-6645158 on Build GRCh38
rs756596238 Chr.12: 6645200-6645200 on Build GRCh38
rs538633343 Chr.12: 6645157-6645157 on Build GRCh38
rs757048452 Chr.12: 6645178-6645178 on Build GRCh38
rs945141392 Chr.12: 6645496-6645496 on Build GRCh38
rs567499982 Chr.12: 6645107-6645107 on Build GRCh38
rs1022809146 Chr.12: 6645337-6645337 on Build GRCh38
rs778495838 Chr.12: 6645230-6645230 on Build GRCh38
rs554829355 Chr.12: 6645250-6645250 on Build GRCh38
rs749599320 Chr.12: 6645208-6645208 on Build GRCh38
rs760697193 Chr.12: 6645259-6645259 on Build GRCh38
rs375082444 Chr.12: 6645251-6645251 on Build GRCh38
rs932317868 Chr.12: 6645412-6645412 on Build GRCh38
rs1053551657 Chr.12: 6645143-6645143 on Build GRCh38
rs35729062 Chr.12: 6645313-6645313 on Build GRCh38
rs545402806 Chr.12: 6645434-6645434 on Build GRCh38
rs768629998 Chr.12: 6645518-6645518 on Build GRCh38
rs1053435702 Chr.12: 6645465-6645465 on Build GRCh38
rs922155015 Chr.12: 6645384-6645384 on Build GRCh38
rs368595038 Chr.12: 6645378-6645378 on Build GRCh38
rs541314219 Chr.12: 6645199-6645199 on Build GRCh38
rs779394139 Chr.12: 6645345-6645345 on Build GRCh38
rs768602472 Chr.12: 6645361-6645361 on Build GRCh38
rs893526626 Chr.12: 6645146-6645146 on Build GRCh38

View Full Product Details