Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147649962-147650387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP6
Entrez Gene ID
51205
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
ACPL1, LPAP, PACPL1
UniGene
Hs.562154
Gene Chromosome Location
Chr.1: 147629658-147670543 on Build GRCh38
Transcripts
NM_001323625.1 NM_016361.4 NR_136633.1 NR_136634.1 NR_136635.1 NR_136636.1 XM_011509601.2 XM_017001422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592439 c.964C>T p.P322S Chr.1: 147650156-147650156 on Build GRCh38
3473208 c.908C>T p.P303L Chr.1: 147650212-147650212 on Build GRCh38
242783 c.894G>A p.Q298Q Chr.1: 147650226-147650226 on Build GRCh38
7206618 c.973A>C p.I325L Chr.1: 147650147-147650147 on Build GRCh38
6373432 c.961G>C p.A321P Chr.1: 147650159-147650159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587678756 Chr.1: 147650007-147650007 on Build GRCh38
rs782307947 Chr.1: 147650308-147650308 on Build GRCh38
rs782648458 Chr.1: 147650142-147650142 on Build GRCh38
rs782192597 Chr.1: 147650158-147650158 on Build GRCh38
rs781962696 Chr.1: 147650199-147650199 on Build GRCh38
rs782225158 Chr.1: 147650256-147650256 on Build GRCh38
rs887884063 Chr.1: 147650293-147650293 on Build GRCh38
rs782593785 Chr.1: 147650249-147650249 on Build GRCh38
rs782410525 Chr.1: 147650193-147650193 on Build GRCh38
rs142610979 Chr.1: 147650102-147650102 on Build GRCh38
rs782600262 Chr.1: 147650111-147650111 on Build GRCh38
rs587689616 Chr.1: 147650353-147650353 on Build GRCh38
rs192281767 Chr.1: 147650195-147650195 on Build GRCh38
rs781788159 Chr.1: 147650241-147650241 on Build GRCh38
rs782514349 Chr.1: 147650262-147650262 on Build GRCh38
rs587595573 Chr.1: 147650009-147650009 on Build GRCh38
rs781991270 Chr.1: 147650171-147650171 on Build GRCh38
rs782805660 Chr.1: 147650234-147650234 on Build GRCh38
rs782341032 Chr.1: 147650163-147650163 on Build GRCh38
rs782530833 Chr.1: 147650231-147650231 on Build GRCh38
rs911125332 Chr.1: 147650148-147650148 on Build GRCh38
rs906125526 Chr.1: 147650360-147650360 on Build GRCh38
rs988529500 Chr.1: 147650050-147650050 on Build GRCh38
rs1044826338 Chr.1: 147650319-147650319 on Build GRCh38
rs1006862298 Chr.1: 147649986-147649986 on Build GRCh38
rs587672456 Chr.1: 147650061-147650061 on Build GRCh38
rs782820624 Chr.1: 147650099-147650099 on Build GRCh38
rs955752492 Chr.1: 147650035-147650035 on Build GRCh38
rs947089529 Chr.1: 147650081-147650081 on Build GRCh38
rs1040895835 Chr.1: 147650186-147650186 on Build GRCh38
rs782576106 Chr.1: 147650154-147650154 on Build GRCh38
rs1047827347 Chr.1: 147650224-147650224 on Build GRCh38
rs6593795 Chr.1: 147650174-147650174 on Build GRCh38
rs981568350 Chr.1: 147650016-147650016 on Build GRCh38
rs782101381 Chr.1: 147650200-147650200 on Build GRCh38
rs377754599 Chr.1: 147650289-147650289 on Build GRCh38
rs181906644 Chr.1: 147650008-147650008 on Build GRCh38
rs375159217 Chr.1: 147650138-147650138 on Build GRCh38
rs781835741 Chr.1: 147650136-147650136 on Build GRCh38
rs782760215 Chr.1: 147650294-147650294 on Build GRCh38
rs1035861376 Chr.1: 147650028-147650028 on Build GRCh38
rs782299810 Chr.1: 147650275-147650275 on Build GRCh38
rs782711818 Chr.1: 147650197-147650197 on Build GRCh38
rs587628195 Chr.1: 147650143-147650143 on Build GRCh38
rs782414489 Chr.1: 147650153-147650153 on Build GRCh38
rs782473324 Chr.1: 147650103-147650103 on Build GRCh38
rs187384544 Chr.1: 147650194-147650194 on Build GRCh38
rs782670059 Chr.1: 147650263-147650263 on Build GRCh38
rs772881894 Chr.1: 147650265-147650265 on Build GRCh38
rs782769484 Chr.1: 147650202-147650202 on Build GRCh38
rs782319455 Chr.1: 147650288-147650288 on Build GRCh38
rs782462247 Chr.1: 147650248-147650248 on Build GRCh38

View Full Product Details