Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99850085-99850651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGL
Entrez Gene ID
178
Gene Name
amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase
Gene Aliases
GDE
UniGene
Hs.904
Gene Chromosome Location
Chr.1: 99850077-99924023 on Build GRCh38
Transcripts
NM_000028.2 NM_000642.2 NM_000643.2 NM_000644.2 XM_005270557.2

SNP Detail

SNP ID Chromosome Location
rs1037419108 Chr.1: 99850438-99850438 on Build GRCh38
rs998615584 Chr.1: 99850445-99850445 on Build GRCh38
rs764383829 Chr.1: 99850445-99850445 on Build GRCh38
rs563389574 Chr.1: 99850276-99850276 on Build GRCh38
rs1046476870 Chr.1: 99850501-99850501 on Build GRCh38
rs1025908351 Chr.1: 99850378-99850378 on Build GRCh38
rs1021151265 Chr.1: 99850563-99850563 on Build GRCh38
rs937844168 Chr.1: 99850389-99850389 on Build GRCh38
rs926565594 Chr.1: 99850383-99850383 on Build GRCh38
rs901189755 Chr.1: 99850442-99850442 on Build GRCh38
rs970401887 Chr.1: 99850598-99850598 on Build GRCh38
rs528586382 Chr.1: 99850299-99850299 on Build GRCh38
rs1000329504 Chr.1: 99850520-99850520 on Build GRCh38
rs982224973 Chr.1: 99850609-99850609 on Build GRCh38
rs1056104580 Chr.1: 99850464-99850464 on Build GRCh38
rs987920790 Chr.1: 99850562-99850562 on Build GRCh38
rs540388561 Chr.1: 99850448-99850449 on Build GRCh38
rs1048445985 Chr.1: 99850170-99850170 on Build GRCh38
rs530809457 Chr.1: 99850526-99850526 on Build GRCh38
rs552648572 Chr.1: 99850433-99850433 on Build GRCh38
rs548243205 Chr.1: 99850350-99850350 on Build GRCh38
rs570733643 Chr.1: 99850626-99850626 on Build GRCh38
rs994296952 Chr.1: 99850371-99850371 on Build GRCh38
rs568150943 Chr.1: 99850468-99850468 on Build GRCh38
rs976887337 Chr.1: 99850275-99850275 on Build GRCh38
rs1046633184 Chr.1: 99850147-99850147 on Build GRCh38
rs948045826 Chr.1: 99850469-99850469 on Build GRCh38
rs959456759 Chr.1: 99850115-99850115 on Build GRCh38
rs1004162902 Chr.1: 99850189-99850189 on Build GRCh38
rs1002022190 Chr.1: 99850156-99850156 on Build GRCh38
rs949298765 Chr.1: 99850355-99850355 on Build GRCh38
rs910974381 Chr.1: 99850215-99850215 on Build GRCh38
rs1053692680 Chr.1: 99850343-99850343 on Build GRCh38
rs983751535 Chr.1: 99850192-99850192 on Build GRCh38
rs954173274 Chr.1: 99850265-99850265 on Build GRCh38
rs1032085337 Chr.1: 99850322-99850322 on Build GRCh38
rs957781051 Chr.1: 99850344-99850344 on Build GRCh38
rs952359111 Chr.1: 99850149-99850149 on Build GRCh38
rs988172683 Chr.1: 99850346-99850346 on Build GRCh38
rs113127962 Chr.1: 99850257-99850257 on Build GRCh38
rs1028883958 Chr.1: 99850262-99850262 on Build GRCh38
rs182466176 Chr.1: 99850196-99850196 on Build GRCh38
rs371657154 Chr.1: 99850596-99850596 on Build GRCh38
rs757978780 Chr.1: 99850242-99850242 on Build GRCh38
rs901384836 Chr.1: 99850209-99850209 on Build GRCh38
rs1048251367 Chr.1: 99850406-99850406 on Build GRCh38
rs998467702 Chr.1: 99850222-99850222 on Build GRCh38
rs1041620474 Chr.1: 99850323-99850323 on Build GRCh38
rs535096205 Chr.1: 99850472-99850472 on Build GRCh38
rs888498974 Chr.1: 99850408-99850408 on Build GRCh38
rs888570692 Chr.1: 99850171-99850171 on Build GRCh38
rs905038754 Chr.1: 99850154-99850154 on Build GRCh38
rs777278669 Chr.1: 99850434-99850434 on Build GRCh38
rs904416881 Chr.1: 99850327-99850327 on Build GRCh38
rs559643679 Chr.1: 99850133-99850133 on Build GRCh38
rs550641188 Chr.1: 99850542-99850542 on Build GRCh38
rs971291214 Chr.1: 99850134-99850134 on Build GRCh38
rs913774174 Chr.1: 99850128-99850128 on Build GRCh38
rs936689377 Chr.1: 99850336-99850336 on Build GRCh38
rs973014607 Chr.1: 99850143-99850143 on Build GRCh38
rs186104051 Chr.1: 99850240-99850240 on Build GRCh38
rs566013647 Chr.1: 99850448-99850448 on Build GRCh38
rs957750361 Chr.1: 99850540-99850540 on Build GRCh38
rs1032398421 Chr.1: 99850109-99850109 on Build GRCh38
rs906740772 Chr.1: 99850356-99850356 on Build GRCh38
rs906575280 Chr.1: 99850522-99850522 on Build GRCh38
rs994349735 Chr.1: 99850527-99850527 on Build GRCh38
rs935945280 Chr.1: 99850458-99850458 on Build GRCh38
rs983805364 Chr.1: 99850400-99850400 on Build GRCh38
rs982037804 Chr.1: 99850380-99850380 on Build GRCh38

View Full Product Details