Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32501211-32501668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVA1C
Entrez Gene ID
59271
Gene Name
eva-1 homolog C (C. elegans)
Gene Aliases
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21
UniGene
Hs.208358
Gene Chromosome Location
Chr.21: 32411692-32515400 on Build GRCh38
Transcripts
NM_001286556.1 NM_001320744.1 NM_001320745.1 NM_058187.4 NR_104472.1 NR_135467.1 XM_005261019.4 XM_006724038.3 XM_011529669.2 XM_017028418.1 XM_017028419.1 XM_017028420.1 XM_017028421.1 XM_017028422.1 XM_017028423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682739 c.792A>T p.I264I Chr.21: 32501428-32501428 on Build GRCh38
3550270 c.848A>C p.D283A Chr.21: 32501484-32501484 on Build GRCh38
159573 c.786G>A p.K262K Chr.21: 32501422-32501422 on Build GRCh38
1030192 c.859+7T>G p.? Chr.21: 32501502-32501502 on Build GRCh38
5779862 c.800C>T p.A267V Chr.21: 32501436-32501436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397744435 Chr.21: 32501249-32501250 on Build GRCh38
rs114508192 Chr.21: 32501634-32501634 on Build GRCh38
rs35132587 Chr.21: 32501248-32501249 on Build GRCh38
rs771236590 Chr.21: 32501573-32501573 on Build GRCh38
rs747707021 Chr.21: 32501423-32501424 on Build GRCh38
rs749667202 Chr.21: 32501572-32501572 on Build GRCh38
rs142626450 Chr.21: 32501436-32501436 on Build GRCh38
rs144869620 Chr.21: 32501563-32501563 on Build GRCh38
rs757928232 Chr.21: 32501450-32501450 on Build GRCh38
rs199920078 Chr.21: 32501250-32501250 on Build GRCh38
rs550292110 Chr.21: 32501384-32501384 on Build GRCh38
rs936346333 Chr.21: 32501636-32501636 on Build GRCh38
rs759546002 Chr.21: 32501419-32501419 on Build GRCh38
rs959156262 Chr.21: 32501605-32501605 on Build GRCh38
rs753687467 Chr.21: 32501523-32501523 on Build GRCh38
rs555268310 Chr.21: 32501622-32501622 on Build GRCh38
rs532326165 Chr.21: 32501438-32501438 on Build GRCh38
rs1053505922 Chr.21: 32501637-32501637 on Build GRCh38
rs748479622 Chr.21: 32501391-32501391 on Build GRCh38
rs561559756 Chr.21: 32501269-32501269 on Build GRCh38
rs746416972 Chr.21: 32501578-32501578 on Build GRCh38
rs779604529 Chr.21: 32501451-32501451 on Build GRCh38
rs746714613 Chr.21: 32501471-32501471 on Build GRCh38
rs763993874 Chr.21: 32501434-32501434 on Build GRCh38
rs200813543 Chr.21: 32501439-32501439 on Build GRCh38
rs201073595 Chr.21: 32501253-32501253 on Build GRCh38
rs775405145 Chr.21: 32501511-32501511 on Build GRCh38
rs749650775 Chr.21: 32501485-32501485 on Build GRCh38
rs8132528 Chr.21: 32501619-32501619 on Build GRCh38
rs565185312 Chr.21: 32501338-32501338 on Build GRCh38
rs779663735 Chr.21: 32501549-32501549 on Build GRCh38
rs770154444 Chr.21: 32501393-32501393 on Build GRCh38
rs773416929 Chr.21: 32501399-32501399 on Build GRCh38
rs766860667 Chr.21: 32501407-32501407 on Build GRCh38
rs771851447 Chr.21: 32501383-32501383 on Build GRCh38
rs761009876 Chr.21: 32501513-32501513 on Build GRCh38
rs745364554 Chr.21: 32501366-32501366 on Build GRCh38
rs773681344 Chr.21: 32501484-32501484 on Build GRCh38
rs767647216 Chr.21: 32501426-32501426 on Build GRCh38
rs771432974 Chr.21: 32501494-32501494 on Build GRCh38
rs745921827 Chr.21: 32501284-32501287 on Build GRCh38
rs571660362 Chr.21: 32501433-32501433 on Build GRCh38
rs764212160 Chr.21: 32501521-32501521 on Build GRCh38
rs1014950709 Chr.21: 32501418-32501418 on Build GRCh38
rs936456550 Chr.21: 32501246-32501246 on Build GRCh38
rs750342159 Chr.21: 32501539-32501539 on Build GRCh38
rs566412995 Chr.21: 32501618-32501618 on Build GRCh38
rs763497663 Chr.21: 32501400-32501400 on Build GRCh38
rs759635568 Chr.21: 32501506-32501506 on Build GRCh38
rs765284411 Chr.21: 32501534-32501534 on Build GRCh38
rs1051911232 Chr.21: 32501647-32501647 on Build GRCh38
rs537652588 Chr.21: 32501644-32501644 on Build GRCh38
rs771692058 Chr.21: 32501504-32501505 on Build GRCh38
rs774482044 Chr.21: 32501408-32501408 on Build GRCh38
rs978141116 Chr.21: 32501633-32501633 on Build GRCh38
rs758237223 Chr.21: 32501545-32501545 on Build GRCh38
rs757057379 Chr.21: 32501525-32501525 on Build GRCh38
rs774782469 Chr.21: 32501500-32501500 on Build GRCh38
rs531709970 Chr.21: 32501291-32501291 on Build GRCh38
rs970659359 Chr.21: 32501632-32501632 on Build GRCh38
rs996308748 Chr.21: 32501336-32501336 on Build GRCh38
rs768169224 Chr.21: 32501474-32501474 on Build GRCh38
rs781071744 Chr.21: 32501608-32501608 on Build GRCh38
rs751018233 Chr.21: 32501552-32501552 on Build GRCh38

View Full Product Details