Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 143945894-143946491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
UniGene
Hs.44780
Gene Chromosome Location
Chr.2: 143945787-144333092 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

Gene Symbol
LOC101928386
Entrez Gene ID
101928386
Gene Name
uncharacterized LOC101928386
Gene Aliases
-
UniGene
Hs.668251
Gene Chromosome Location
Chr.2: 143937067-143964156 on Build GRCh38
Transcripts
NM_001006636.3 NM_001164629.3 NM_001284233.1 NM_001284234.1 NM_001284238.1 NM_024659.4 XM_005263774.2 XM_005263782.2 XM_005263784.2 XM_011511843.2 XM_011511855.2 XM_011511856.2 XM_017004917.1 XM_017004918.1 XM_017004919.1 XM_017004920.1 XM_017004921.1 XM_017004922.1 XM_017004923.1 XM_017004924.1 XM_017004925.1 XM_017004926.1 XM_017004927.1 XM_017004928.1 XM_017004929.1 XM_017004930.1 XM_017004931.1 XM_017004932.1 XM_017004933.1 XM_017004934.1 XM_017004935.1 XM_017004936.1 XM_017004937.1 XM_017004938.1 XM_017004939.1 XM_017004940.1 XM_017004941.1 XM_017004942.1 XM_017004943.1 XM_017004944.1 XM_017004945.1 XM_017004946.1 XM_017004947.1 XM_017004948.1 XM_017004949.1 XM_017004950.1 XM_017004951.1 XM_017004952.1 XM_017004953.1 XM_017004954.1 XM_017004955.1 XM_017004956.1 XM_017004957.1 NR_110237.1

SNP Detail

SNP ID Chromosome Location
rs940198529 Chr.2: 143946376-143946376 on Build GRCh38
rs909946241 Chr.2: 143946340-143946343 on Build GRCh38
rs893149869 Chr.2: 143945982-143945982 on Build GRCh38
rs866159076 Chr.2: 143946229-143946229 on Build GRCh38
rs950018974 Chr.2: 143946324-143946324 on Build GRCh38
rs910067850 Chr.2: 143946003-143946003 on Build GRCh38
rs987021213 Chr.2: 143946213-143946213 on Build GRCh38
rs762822640 Chr.2: 143946331-143946331 on Build GRCh38
rs955725008 Chr.2: 143946133-143946133 on Build GRCh38
rs1046227341 Chr.2: 143945941-143945941 on Build GRCh38
rs1001988654 Chr.2: 143946147-143946147 on Build GRCh38
rs571404699 Chr.2: 143946047-143946047 on Build GRCh38
rs182585056 Chr.2: 143946452-143946452 on Build GRCh38
rs761535240 Chr.2: 143946364-143946365 on Build GRCh38
rs997039931 Chr.2: 143946108-143946108 on Build GRCh38
rs772182677 Chr.2: 143945943-143945943 on Build GRCh38
rs116007755 Chr.2: 143946384-143946384 on Build GRCh38
rs1028555576 Chr.2: 143946119-143946119 on Build GRCh38
rs552693598 Chr.2: 143945957-143945957 on Build GRCh38
rs1028985459 Chr.2: 143946449-143946449 on Build GRCh38
rs201238279 Chr.2: 143945924-143945924 on Build GRCh38
rs984212186 Chr.2: 143945988-143945988 on Build GRCh38
rs769802549 Chr.2: 143946300-143946300 on Build GRCh38
rs538732062 Chr.2: 143946054-143946054 on Build GRCh38
rs776917212 Chr.2: 143946137-143946140 on Build GRCh38
rs189026966 Chr.2: 143946308-143946308 on Build GRCh38
rs950320384 Chr.2: 143946426-143946426 on Build GRCh38
rs181676446 Chr.2: 143946163-143946163 on Build GRCh38
rs1037710595 Chr.2: 143946081-143946081 on Build GRCh38
rs573044687 Chr.2: 143946373-143946373 on Build GRCh38
rs185916563 Chr.2: 143946283-143946283 on Build GRCh38
rs917509324 Chr.2: 143946418-143946418 on Build GRCh38
rs893648899 Chr.2: 143946191-143946191 on Build GRCh38
rs893120183 Chr.2: 143946397-143946397 on Build GRCh38
rs940186942 Chr.2: 143946039-143946040 on Build GRCh38
rs999345468 Chr.2: 143945956-143945956 on Build GRCh38
rs903031462 Chr.2: 143946340-143946340 on Build GRCh38
rs759874721 Chr.2: 143945973-143945973 on Build GRCh38
rs527903328 Chr.2: 143945934-143945934 on Build GRCh38
rs776531896 Chr.2: 143946239-143946239 on Build GRCh38
rs557021934 Chr.2: 143946092-143946092 on Build GRCh38
rs965361638 Chr.2: 143945925-143945925 on Build GRCh38
rs961797868 Chr.2: 143946236-143946236 on Build GRCh38
rs996842048 Chr.2: 143946446-143946446 on Build GRCh38
rs787174 Chr.2: 143945933-143945933 on Build GRCh38
rs934715880 Chr.2: 143946345-143946345 on Build GRCh38
rs930320607 Chr.2: 143946256-143946256 on Build GRCh38
rs1032072865 Chr.2: 143946149-143946149 on Build GRCh38
rs1016184913 Chr.2: 143946228-143946228 on Build GRCh38
rs1032948767 Chr.2: 143945978-143945978 on Build GRCh38
rs949042008 Chr.2: 143946130-143946131 on Build GRCh38
rs1044661461 Chr.2: 143946469-143946469 on Build GRCh38
rs971821594 Chr.2: 143946238-143946238 on Build GRCh38

View Full Product Details