Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 214931879-214932467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA12
Entrez Gene ID
26154
Gene Name
ATP binding cassette subfamily A member 12
Gene Aliases
ARCI4A, ARCI4B, ICR2B, LI2
UniGene
Hs.134585
Gene Chromosome Location
Chr.2: 214931542-215138591 on Build GRCh38
Transcripts
NM_015657.3 NM_173076.2 NR_103740.1 XM_011510951.2 NR_110292.1

Gene Symbol
LOC101928103
Entrez Gene ID
101928103
Gene Name
uncharacterized LOC101928103
Gene Aliases
-
UniGene
Hs.665619
Gene Chromosome Location
Chr.2: 214810229-214963605 on Build GRCh38
Transcripts
NM_015657.3 NM_173076.2 NR_103740.1 XM_011510951.2 NR_110292.1

SNP Detail

SNP ID Chromosome Location
rs567012898 Chr.2: 214932199-214932201 on Build GRCh38
rs936995273 Chr.2: 214932048-214932048 on Build GRCh38
rs139753893 Chr.2: 214932094-214932095 on Build GRCh38
rs896963906 Chr.2: 214932006-214932006 on Build GRCh38
rs763395587 Chr.2: 214932059-214932059 on Build GRCh38
rs186627930 Chr.2: 214931944-214931944 on Build GRCh38
rs979302149 Chr.2: 214932228-214932228 on Build GRCh38
rs180771753 Chr.2: 214931914-214931914 on Build GRCh38
rs1006582201 Chr.2: 214932340-214932340 on Build GRCh38
rs1047361893 Chr.2: 214932067-214932067 on Build GRCh38
rs77729645 Chr.2: 214932149-214932149 on Build GRCh38
rs563265817 Chr.2: 214932054-214932054 on Build GRCh38
rs866709221 Chr.2: 214932083-214932083 on Build GRCh38
rs966295315 Chr.2: 214932118-214932118 on Build GRCh38
rs926203339 Chr.2: 214932254-214932254 on Build GRCh38
rs1055128834 Chr.2: 214931947-214931947 on Build GRCh38
rs535404027 Chr.2: 214932109-214932110 on Build GRCh38
rs529492635 Chr.2: 214932274-214932274 on Build GRCh38
rs1009273219 Chr.2: 214932244-214932244 on Build GRCh38
rs979022898 Chr.2: 214931996-214931996 on Build GRCh38
rs954974886 Chr.2: 214932176-214932176 on Build GRCh38
rs976330417 Chr.2: 214932137-214932137 on Build GRCh38
rs908409878 Chr.2: 214932362-214932362 on Build GRCh38
rs1040700426 Chr.2: 214932344-214932344 on Build GRCh38
rs191323693 Chr.2: 214931970-214931970 on Build GRCh38
rs949346572 Chr.2: 214932238-214932238 on Build GRCh38
rs750719136 Chr.2: 214932269-214932269 on Build GRCh38
rs139852425 Chr.2: 214932338-214932338 on Build GRCh38
rs916901092 Chr.2: 214932306-214932306 on Build GRCh38
rs941205929 Chr.2: 214932380-214932380 on Build GRCh38
rs1036912279 Chr.2: 214932264-214932268 on Build GRCh38
rs774136497 Chr.2: 214932337-214932338 on Build GRCh38
rs931197666 Chr.2: 214932284-214932284 on Build GRCh38
rs996140169 Chr.2: 214932169-214932169 on Build GRCh38
rs751602781 Chr.2: 214932096-214932096 on Build GRCh38
rs925274971 Chr.2: 214931919-214931919 on Build GRCh38
rs540779182 Chr.2: 214931903-214931903 on Build GRCh38
rs183185732 Chr.2: 214932082-214932082 on Build GRCh38
rs1020899336 Chr.2: 214932189-214932189 on Build GRCh38
rs749021141 Chr.2: 214932315-214932318 on Build GRCh38
rs899773224 Chr.2: 214932407-214932407 on Build GRCh38
rs900810732 Chr.2: 214932366-214932366 on Build GRCh38
rs925649742 Chr.2: 214932000-214932000 on Build GRCh38
rs1021171196 Chr.2: 214932420-214932420 on Build GRCh38
rs530493676 Chr.2: 214932060-214932060 on Build GRCh38
rs938060518 Chr.2: 214932218-214932218 on Build GRCh38
rs368041060 Chr.2: 214932324-214932324 on Build GRCh38
rs937959084 Chr.2: 214931933-214931933 on Build GRCh38
rs568623121 Chr.2: 214932170-214932170 on Build GRCh38
rs991171235 Chr.2: 214932287-214932287 on Build GRCh38
rs774394076 Chr.2: 214932399-214932399 on Build GRCh38
rs996693031 Chr.2: 214932422-214932422 on Build GRCh38
rs1038134303 Chr.2: 214932389-214932389 on Build GRCh38
rs917897549 Chr.2: 214932237-214932237 on Build GRCh38
rs879722109 Chr.2: 214932179-214932179 on Build GRCh38
rs147463711 Chr.2: 214931902-214931902 on Build GRCh38
rs547358293 Chr.2: 214932161-214932161 on Build GRCh38
rs995667824 Chr.2: 214932037-214932037 on Build GRCh38
rs1050407481 Chr.2: 214932426-214932426 on Build GRCh38
rs886055603 Chr.2: 214932238-214932238 on Build GRCh38
rs1046805512 Chr.2: 214932359-214932359 on Build GRCh38
rs372365280 Chr.2: 214932164-214932164 on Build GRCh38
rs988056388 Chr.2: 214931983-214931983 on Build GRCh38
rs967570879 Chr.2: 214932223-214932223 on Build GRCh38
rs555497112 Chr.2: 214931988-214931988 on Build GRCh38
rs1048211793 Chr.2: 214932314-214932315 on Build GRCh38
rs998177251 Chr.2: 214932403-214932403 on Build GRCh38
rs189866805 Chr.2: 214931901-214931901 on Build GRCh38
rs886055604 Chr.2: 214932256-214932256 on Build GRCh38
rs1029036546 Chr.2: 214931962-214931962 on Build GRCh38
rs572013028 Chr.2: 214932123-214932123 on Build GRCh38
rs564726033 Chr.2: 214932120-214932120 on Build GRCh38
rs570560109 Chr.2: 214932336-214932340 on Build GRCh38
rs574584043 Chr.2: 214931940-214931940 on Build GRCh38
rs946312884 Chr.2: 214931993-214931993 on Build GRCh38
rs367714646 Chr.2: 214932178-214932178 on Build GRCh38
rs1019220416 Chr.2: 214932095-214932095 on Build GRCh38

View Full Product Details