Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1004592-1005187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABR
Entrez Gene ID
29
Gene Name
active BCR-related
Gene Aliases
MDB
UniGene
Hs.159306
Gene Chromosome Location
Chr.17: 1003518-1229713 on Build GRCh38
Transcripts
NM_001092.4 NM_001159746.2 NM_001256847.2 NM_001282149.1 NM_001322840.1 NM_001322841.1 NM_021962.4 XM_011523810.2 XM_011523812.2 XM_011523813.2 XM_011523814.2 XM_011523815.2 XM_017024539.1 XM_017024540.1 XM_017024542.1 XM_017024543.1

SNP Detail

SNP ID Chromosome Location
rs530843257 Chr.17: 1004673-1004673 on Build GRCh38
rs1024546846 Chr.17: 1004949-1004949 on Build GRCh38
rs200536245 Chr.17: 1005079-1005079 on Build GRCh38
rs908936643 Chr.17: 1004845-1004845 on Build GRCh38
rs867338376 Chr.17: 1005071-1005071 on Build GRCh38
rs963081100 Chr.17: 1005073-1005073 on Build GRCh38
rs536219816 Chr.17: 1004951-1004951 on Build GRCh38
rs767199986 Chr.17: 1005078-1005084 on Build GRCh38
rs181765877 Chr.17: 1004988-1004988 on Build GRCh38
rs951483120 Chr.17: 1004985-1004985 on Build GRCh38
rs1007279653 Chr.17: 1004935-1004935 on Build GRCh38
rs937760853 Chr.17: 1005088-1005088 on Build GRCh38
rs919092084 Chr.17: 1004875-1004875 on Build GRCh38
rs909951647 Chr.17: 1004798-1004798 on Build GRCh38
rs1018736503 Chr.17: 1004710-1004710 on Build GRCh38
rs1014931492 Chr.17: 1004939-1004939 on Build GRCh38
rs1009746336 Chr.17: 1004611-1004611 on Build GRCh38
rs748824261 Chr.17: 1004646-1004646 on Build GRCh38
rs979541502 Chr.17: 1004727-1004727 on Build GRCh38
rs550721736 Chr.17: 1004937-1004937 on Build GRCh38
rs918208359 Chr.17: 1004781-1004781 on Build GRCh38
rs935598624 Chr.17: 1004615-1004615 on Build GRCh38
rs3744753 Chr.17: 1004667-1004667 on Build GRCh38
rs147858840 Chr.17: 1004647-1004647 on Build GRCh38
rs992451930 Chr.17: 1004621-1004621 on Build GRCh38
rs921068344 Chr.17: 1004848-1004848 on Build GRCh38
rs921110491 Chr.17: 1005021-1005021 on Build GRCh38
rs769118358 Chr.17: 1004994-1004994 on Build GRCh38
rs776752704 Chr.17: 1005015-1005015 on Build GRCh38
rs776186525 Chr.17: 1004792-1004792 on Build GRCh38
rs546728923 Chr.17: 1004877-1004877 on Build GRCh38
rs773798891 Chr.17: 1004973-1004975 on Build GRCh38
rs756099387 Chr.17: 1004614-1004614 on Build GRCh38
rs775780189 Chr.17: 1004929-1004929 on Build GRCh38
rs535052489 Chr.17: 1004999-1004999 on Build GRCh38
rs542637327 Chr.17: 1004737-1004737 on Build GRCh38
rs968898980 Chr.17: 1004627-1004627 on Build GRCh38
rs772452385 Chr.17: 1004905-1004905 on Build GRCh38
rs968123292 Chr.17: 1004726-1004726 on Build GRCh38
rs3744755 Chr.17: 1004989-1004989 on Build GRCh38
rs333642 Chr.17: 1004818-1004818 on Build GRCh38
rs118178620 Chr.17: 1005112-1005112 on Build GRCh38
rs190121096 Chr.17: 1004940-1004940 on Build GRCh38
rs577852586 Chr.17: 1005078-1005078 on Build GRCh38
rs894661442 Chr.17: 1005094-1005094 on Build GRCh38
rs557009495 Chr.17: 1005091-1005091 on Build GRCh38
rs971801875 Chr.17: 1005074-1005074 on Build GRCh38
rs1006901246 Chr.17: 1004709-1004709 on Build GRCh38
rs1000363796 Chr.17: 1004638-1004638 on Build GRCh38
rs982858454 Chr.17: 1004778-1004778 on Build GRCh38
rs529767240 Chr.17: 1004817-1004817 on Build GRCh38
rs552189489 Chr.17: 1005146-1005146 on Build GRCh38
rs1031114932 Chr.17: 1004740-1004743 on Build GRCh38
rs778641157 Chr.17: 1004752-1004752 on Build GRCh38
rs894618241 Chr.17: 1004930-1004930 on Build GRCh38
rs933920180 Chr.17: 1004862-1004862 on Build GRCh38
rs368338298 Chr.17: 1005096-1005096 on Build GRCh38
rs575351787 Chr.17: 1005110-1005110 on Build GRCh38
rs997827157 Chr.17: 1004685-1004685 on Build GRCh38
rs891114596 Chr.17: 1004696-1004696 on Build GRCh38
rs1000855014 Chr.17: 1005127-1005127 on Build GRCh38
rs3744754 Chr.17: 1004903-1004903 on Build GRCh38
rs1050995091 Chr.17: 1004865-1004865 on Build GRCh38
rs943287649 Chr.17: 1005108-1005108 on Build GRCh38
rs528385293 Chr.17: 1004840-1004840 on Build GRCh38
rs185093674 Chr.17: 1004642-1004642 on Build GRCh38
rs201385837 Chr.17: 1005147-1005148 on Build GRCh38
rs1014145795 Chr.17: 1005162-1005162 on Build GRCh38
rs372164467 Chr.17: 1005111-1005111 on Build GRCh38
rs887358747 Chr.17: 1005126-1005126 on Build GRCh38
rs746614823 Chr.17: 1004665-1004666 on Build GRCh38
rs777467640 Chr.17: 1004625-1004625 on Build GRCh38
rs538727329 Chr.17: 1005090-1005090 on Build GRCh38
rs903598609 Chr.17: 1004907-1004907 on Build GRCh38
rs987148512 Chr.17: 1005059-1005059 on Build GRCh38
rs754443572 Chr.17: 1004803-1004803 on Build GRCh38
rs759169646 Chr.17: 1005047-1005049 on Build GRCh38
rs867642766 Chr.17: 1005086-1005086 on Build GRCh38
rs562241396 Chr.17: 1004684-1004684 on Build GRCh38
rs1030289484 Chr.17: 1004691-1004691 on Build GRCh38
rs571399770 Chr.17: 1004878-1004878 on Build GRCh38
rs553521155 Chr.17: 1005004-1005004 on Build GRCh38
rs747777019 Chr.17: 1004997-1004997 on Build GRCh38
rs962771786 Chr.17: 1005008-1005008 on Build GRCh38
rs1001918416 Chr.17: 1004923-1004923 on Build GRCh38
rs1004384008 Chr.17: 1005131-1005131 on Build GRCh38
rs770114929 Chr.17: 1004880-1004880 on Build GRCh38
rs945059762 Chr.17: 1005072-1005072 on Build GRCh38
rs962657327 Chr.17: 1005147-1005147 on Build GRCh38
rs933827453 Chr.17: 1005035-1005035 on Build GRCh38
rs1025606473 Chr.17: 1005003-1005003 on Build GRCh38
rs761968267 Chr.17: 1005062-1005062 on Build GRCh38
rs757973313 Chr.17: 1005074-1005080 on Build GRCh38

View Full Product Details