Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 9204239-9204765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD12
Entrez Gene ID
23253
Gene Name
ankyrin repeat domain 12
Gene Aliases
ANCO-2, ANCO1, GAC-1, Nbla00144
UniGene
Hs.464585
Gene Chromosome Location
Chr.18: 9136753-9285985 on Build GRCh38
Transcripts
NM_001083625.2 NM_001204056.1 NM_015208.4 XM_005258092.3 XM_005258093.3 XM_005258095.2 XM_005258096.2 XM_011525638.2 XM_011525639.2 XM_017025661.1 XM_017025662.1 XM_017025663.1 XM_017025664.1 XM_017025665.1 XM_017025666.1 XM_017025667.1 XM_017025668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4073647 c.288T>G p.S96S Chr.18: 9204528-9204528 on Build GRCh38
5570505 c.295A>C p.T99P Chr.18: 9204535-9204535 on Build GRCh38
6600356 c.299A>C p.Y100S Chr.18: 9204539-9204539 on Build GRCh38
5056160 c.304+2T>C p.? Chr.18: 9204546-9204546 on Build GRCh38
3701406 c.267G>A p.W89* Chr.18: 9204507-9204507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576346090 Chr.18: 9204302-9204302 on Build GRCh38
rs747136144 Chr.18: 9204452-9204454 on Build GRCh38
rs1001399675 Chr.18: 9204329-9204329 on Build GRCh38
rs1001664203 Chr.18: 9204671-9204671 on Build GRCh38
rs762515175 Chr.18: 9204455-9204455 on Build GRCh38
rs758540298 Chr.18: 9204510-9204510 on Build GRCh38
rs775536238 Chr.18: 9204440-9204441 on Build GRCh38
rs768600031 Chr.18: 9204539-9204542 on Build GRCh38
rs754371932 Chr.18: 9204504-9204504 on Build GRCh38
rs993237365 Chr.18: 9204277-9204277 on Build GRCh38
rs780095195 Chr.18: 9204529-9204529 on Build GRCh38
rs766838475 Chr.18: 9204496-9204496 on Build GRCh38
rs778977438 Chr.18: 9204507-9204507 on Build GRCh38
rs769698224 Chr.18: 9204425-9204425 on Build GRCh38
rs968645199 Chr.18: 9204381-9204381 on Build GRCh38
rs745622553 Chr.18: 9204516-9204516 on Build GRCh38
rs750742251 Chr.18: 9204492-9204492 on Build GRCh38
rs1011507222 Chr.18: 9204685-9204685 on Build GRCh38
rs775654324 Chr.18: 9204459-9204459 on Build GRCh38
rs906981248 Chr.18: 9204609-9204609 on Build GRCh38
rs1034583745 Chr.18: 9204356-9204356 on Build GRCh38
rs1056934003 Chr.18: 9204659-9204659 on Build GRCh38
rs142771790 Chr.18: 9204521-9204521 on Build GRCh38
rs774000988 Chr.18: 9204464-9204464 on Build GRCh38
rs1012578494 Chr.18: 9204307-9204307 on Build GRCh38
rs531205137 Chr.18: 9204725-9204725 on Build GRCh38
rs183131931 Chr.18: 9204413-9204413 on Build GRCh38
rs201125084 Chr.18: 9204435-9204435 on Build GRCh38
rs768215108 Chr.18: 9204550-9204550 on Build GRCh38
rs756521562 Chr.18: 9204494-9204494 on Build GRCh38
rs80347186 Chr.18: 9204738-9204738 on Build GRCh38
rs906946501 Chr.18: 9204309-9204309 on Build GRCh38
rs1056821497 Chr.18: 9204281-9204281 on Build GRCh38
rs1022925912 Chr.18: 9204358-9204358 on Build GRCh38
rs530338120 Chr.18: 9204561-9204561 on Build GRCh38
rs536323009 Chr.18: 9204290-9204290 on Build GRCh38
rs886068764 Chr.18: 9204291-9204291 on Build GRCh38
rs1045793913 Chr.18: 9204606-9204606 on Build GRCh38
rs897581771 Chr.18: 9204275-9204275 on Build GRCh38
rs1000467925 Chr.18: 9204412-9204412 on Build GRCh38
rs142497652 Chr.18: 9204556-9204556 on Build GRCh38
rs919393606 Chr.18: 9204578-9204578 on Build GRCh38
rs761378077 Chr.18: 9204484-9204484 on Build GRCh38
rs929047408 Chr.18: 9204621-9204621 on Build GRCh38
rs1045521457 Chr.18: 9204296-9204296 on Build GRCh38
rs528312252 Chr.18: 9204663-9204663 on Build GRCh38
rs772956994 Chr.18: 9204564-9204564 on Build GRCh38
rs561072075 Chr.18: 9204676-9204676 on Build GRCh38
rs140251903 Chr.18: 9204293-9204293 on Build GRCh38
rs745739761 Chr.18: 9204434-9204434 on Build GRCh38
rs779819143 Chr.18: 9204429-9204429 on Build GRCh38
rs778112050 Chr.18: 9204513-9204513 on Build GRCh38
rs543836790 Chr.18: 9204352-9204352 on Build GRCh38
rs910820407 Chr.18: 9204522-9204522 on Build GRCh38
rs370484988 Chr.18: 9204672-9204672 on Build GRCh38
rs1012611042 Chr.18: 9204710-9204710 on Build GRCh38
rs1037859572 Chr.18: 9204268-9204268 on Build GRCh38
rs112568144 Chr.18: 9204537-9204537 on Build GRCh38
rs767266765 Chr.18: 9204490-9204490 on Build GRCh38

View Full Product Details