Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 2934947-2935396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSE
Entrez Gene ID
415
Gene Name
arylsulfatase E (chondrodysplasia punctata 1)
Gene Aliases
ASE, CDPX, CDPX1, CDPXR
UniGene
Hs.386975
Gene Chromosome Location
Chr.X: 2934632-2968310 on Build GRCh38
Transcripts
NM_000047.2 NM_001282628.1 NM_001282631.1 XM_005274518.2 XM_005274519.4 XM_005274521.4 XM_011545521.1 XM_017029525.1 XM_017029526.1

SNP Detail

SNP ID Chromosome Location
rs957923385 Chr.X: 2935313-2935313 on Build GRCh38
rs762393900 Chr.X: 2935134-2935134 on Build GRCh38
rs762734231 Chr.X: 2935078-2935078 on Build GRCh38
rs369409927 Chr.X: 2934983-2934983 on Build GRCh38
rs751120737 Chr.X: 2935248-2935248 on Build GRCh38
rs80338713 Chr.X: 2935160-2935160 on Build GRCh38
rs122460155 Chr.X: 2935127-2935127 on Build GRCh38
rs747510606 Chr.X: 2935021-2935021 on Build GRCh38
rs769363825 Chr.X: 2935212-2935212 on Build GRCh38
rs757422760 Chr.X: 2935136-2935136 on Build GRCh38
rs755373707 Chr.X: 2935183-2935183 on Build GRCh38
rs766206289 Chr.X: 2935089-2935089 on Build GRCh38
rs780330895 Chr.X: 2935159-2935159 on Build GRCh38
rs772699055 Chr.X: 2935123-2935123 on Build GRCh38
rs758804487 Chr.X: 2935156-2935156 on Build GRCh38
rs143775522 Chr.X: 2935184-2935184 on Build GRCh38
rs759279080 Chr.X: 2935266-2935266 on Build GRCh38
rs75821383 Chr.X: 2935105-2935105 on Build GRCh38
rs767739303 Chr.X: 2935298-2935298 on Build GRCh38
rs1011406499 Chr.X: 2935336-2935336 on Build GRCh38
rs376111412 Chr.X: 2935005-2935005 on Build GRCh38
rs139173612 Chr.X: 2934977-2934977 on Build GRCh38
rs993209695 Chr.X: 2935141-2935141 on Build GRCh38
rs756191732 Chr.X: 2934971-2934971 on Build GRCh38
rs765542263 Chr.X: 2935153-2935153 on Build GRCh38
rs768189016 Chr.X: 2935030-2935030 on Build GRCh38
rs753170566 Chr.X: 2935310-2935310 on Build GRCh38
rs749592875 Chr.X: 2935370-2935370 on Build GRCh38
rs753971115 Chr.X: 2934973-2934973 on Build GRCh38
rs775965159 Chr.X: 2935048-2935048 on Build GRCh38
rs772821867 Chr.X: 2935231-2935231 on Build GRCh38
rs750738958 Chr.X: 2935155-2935155 on Build GRCh38
rs780700222 Chr.X: 2935009-2935009 on Build GRCh38
rs747645218 Chr.X: 2935199-2935199 on Build GRCh38
rs747699123 Chr.X: 2935052-2935052 on Build GRCh38
rs772908873 Chr.X: 2935077-2935077 on Build GRCh38
rs756673695 Chr.X: 2935356-2935356 on Build GRCh38
rs141712265 Chr.X: 2935034-2935034 on Build GRCh38
rs769380257 Chr.X: 2935063-2935063 on Build GRCh38
rs55974713 Chr.X: 2935242-2935242 on Build GRCh38
rs758889321 Chr.X: 2935006-2935006 on Build GRCh38
rs752401423 Chr.X: 2934969-2934969 on Build GRCh38
rs371121114 Chr.X: 2935122-2935122 on Build GRCh38
rs41308353 Chr.X: 2935171-2935171 on Build GRCh38
rs751521450 Chr.X: 2934970-2934970 on Build GRCh38
rs756280856 Chr.X: 2935255-2935255 on Build GRCh38
rs967204998 Chr.X: 2935371-2935371 on Build GRCh38
rs1007447288 Chr.X: 2935285-2935285 on Build GRCh38

View Full Product Details