Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107628098-107628662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs886554805 Chr.X: 107628631-107628631 on Build GRCh38
rs913688635 Chr.X: 107628339-107628339 on Build GRCh38
rs1033148112 Chr.X: 107628267-107628267 on Build GRCh38
rs750915295 Chr.X: 107628430-107628430 on Build GRCh38
rs996845821 Chr.X: 107628241-107628241 on Build GRCh38
rs1047821798 Chr.X: 107628221-107628221 on Build GRCh38
rs886126352 Chr.X: 107628253-107628253 on Build GRCh38
rs955158486 Chr.X: 107628367-107628367 on Build GRCh38
rs886438762 Chr.X: 107628167-107628167 on Build GRCh38
rs1044308926 Chr.X: 107628184-107628184 on Build GRCh38
rs1005890415 Chr.X: 107628270-107628270 on Build GRCh38
rs752590804 Chr.X: 107628640-107628640 on Build GRCh38
rs1030564432 Chr.X: 107628364-107628364 on Build GRCh38
rs199968422 Chr.X: 107628556-107628556 on Build GRCh38
rs916698071 Chr.X: 107628534-107628534 on Build GRCh38
rs989529177 Chr.X: 107628526-107628526 on Build GRCh38
rs765785268 Chr.X: 107628291-107628291 on Build GRCh38
rs758827912 Chr.X: 107628454-107628454 on Build GRCh38
rs920387457 Chr.X: 107628539-107628539 on Build GRCh38
rs761523210 Chr.X: 107628585-107628585 on Build GRCh38
rs768856537 Chr.X: 107628476-107628476 on Build GRCh38
rs140689870 Chr.X: 107628216-107628216 on Build GRCh38
rs1047795093 Chr.X: 107628624-107628624 on Build GRCh38
rs957528937 Chr.X: 107628268-107628268 on Build GRCh38
rs991150282 Chr.X: 107628506-107628506 on Build GRCh38
rs371006312 Chr.X: 107628593-107628593 on Build GRCh38
rs944685055 Chr.X: 107628633-107628633 on Build GRCh38
rs958026624 Chr.X: 107628510-107628510 on Build GRCh38
rs978379537 Chr.X: 107628384-107628384 on Build GRCh38
rs981826566 Chr.X: 107628360-107628360 on Build GRCh38
rs964967347 Chr.X: 107628238-107628238 on Build GRCh38
rs948531147 Chr.X: 107628541-107628541 on Build GRCh38
rs1008933786 Chr.X: 107628197-107628197 on Build GRCh38
rs996788623 Chr.X: 107628279-107628279 on Build GRCh38
rs772406932 Chr.X: 107628579-107628579 on Build GRCh38
rs776399830 Chr.X: 107628596-107628596 on Build GRCh38
rs751262798 Chr.X: 107628553-107628553 on Build GRCh38
rs188174974 Chr.X: 107628271-107628271 on Build GRCh38
rs937911805 Chr.X: 107628460-107628460 on Build GRCh38
rs765129709 Chr.X: 107628625-107628625 on Build GRCh38
rs966355978 Chr.X: 107628346-107628346 on Build GRCh38

View Full Product Details