Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA4
Entrez Gene ID
762
Gene Name
carbonic anhydrase 4
Gene Aliases
CAIV, Car4, RP17
Chromosome Location
Chr.17: 60149941 - 60178118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005257639.2 XP_005257696.1 1 - 2 153 109
GenBank mRNA AK289715.1 - 1 - 2 152 109
AK298710.1 - 1 - 2 152 109
BC057792.1 - 1 - 2 158 109
BC069649.1 - 1 - 2 105 109
BC074768.2 - 1 - 2 82 109
DA113846.1 - 1 - 2 153 109
M83670.1 - 1 - 2 106 109

View Full Product Details