The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001807.4 | NP_001798.2 | 5 - 6 | 699 | 55 |
GenBank mRNA | AK299965.1 | - | 5 - 6 | 576 | 55 |
BC042510.1 | - | 5 - 6 | 684 | 55 | |
M85201.1 | - | 5 - 6 | 691 | 55 | |
S79774.1 | - | 5 - 6 | 693 | 55 | |
X54457.1 | - | 5 - 6 | 764 | 55 |