Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179620634-179621164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPH1
Entrez Gene ID
3187
Gene Name
heterogeneous nuclear ribonucleoprotein H1 (H)
Gene Aliases
HNRPH, HNRPH1, hnRNPH
UniGene
Hs.604001
Gene Chromosome Location
Chr.5: 179614178-179634784 on Build GRCh38
Transcripts
NM_001257293.1 NM_005520.2 XM_005265895.2 XM_005265896.2 XM_005265901.1 XM_005265902.4 XM_006714862.2 XM_006714863.2 XM_011534542.1 XM_011534544.1 XM_011534545.1 XM_011534546.1 XM_011534547.1 XM_017009414.1 XM_017009415.1 XM_017009416.1 XM_017009417.1 XM_017009418.1 XM_017009419.1 XM_017009420.1 XM_017009421.1 XM_017009422.1 XM_017009423.1 XM_017009424.1 XM_017009425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6236699 c.263C>A p.S88* Chr.5: 179621026-179621026 on Build GRCh38
6365007 c.365G>T p.C122F Chr.5: 179620924-179620924 on Build GRCh38
212928 c.317A>G p.D106G Chr.5: 179620972-179620972 on Build GRCh38
3011618 c.254-2A>T p.? Chr.5: 179621037-179621037 on Build GRCh38
3011609 c.321G>A p.T107T Chr.5: 179620968-179620968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010351895 Chr.5: 179620827-179620827 on Build GRCh38
rs975716543 Chr.5: 179620671-179620671 on Build GRCh38
rs957801417 Chr.5: 179620813-179620813 on Build GRCh38
rs931304321 Chr.5: 179620952-179620952 on Build GRCh38
rs902647518 Chr.5: 179621120-179621120 on Build GRCh38
rs1023296021 Chr.5: 179620802-179620802 on Build GRCh38
rs1019811514 Chr.5: 179620774-179620774 on Build GRCh38
rs866699842 Chr.5: 179620711-179620711 on Build GRCh38
rs372649727 Chr.5: 179621052-179621052 on Build GRCh38
rs769350421 Chr.5: 179620879-179620879 on Build GRCh38
rs747009222 Chr.5: 179620860-179620860 on Build GRCh38
rs999694640 Chr.5: 179621128-179621128 on Build GRCh38
rs528677343 Chr.5: 179620767-179620767 on Build GRCh38
rs1036310727 Chr.5: 179620756-179620756 on Build GRCh38
rs369373817 Chr.5: 179621067-179621067 on Build GRCh38
rs749005424 Chr.5: 179621076-179621076 on Build GRCh38
rs796813132 Chr.5: 179620666-179620666 on Build GRCh38
rs764677473 Chr.5: 179621005-179621005 on Build GRCh38
rs1041704784 Chr.5: 179621093-179621093 on Build GRCh38
rs761329705 Chr.5: 179620990-179620990 on Build GRCh38
rs956985560 Chr.5: 179620798-179620798 on Build GRCh38
rs758609696 Chr.5: 179621045-179621045 on Build GRCh38
rs909017431 Chr.5: 179620733-179620733 on Build GRCh38
rs1050392634 Chr.5: 179620668-179620668 on Build GRCh38
rs561441353 Chr.5: 179620760-179620760 on Build GRCh38
rs928446533 Chr.5: 179620870-179620870 on Build GRCh38
rs1031432063 Chr.5: 179620796-179620796 on Build GRCh38
rs966224707 Chr.5: 179620873-179620873 on Build GRCh38
rs574160362 Chr.5: 179621044-179621044 on Build GRCh38
rs753096236 Chr.5: 179620969-179620969 on Build GRCh38
rs776047762 Chr.5: 179621084-179621084 on Build GRCh38
rs369999545 Chr.5: 179621050-179621050 on Build GRCh38
rs970028917 Chr.5: 179620833-179620833 on Build GRCh38
rs372880757 Chr.5: 179620850-179620850 on Build GRCh38
rs114883699 Chr.5: 179620780-179620780 on Build GRCh38
rs953172568 Chr.5: 179620971-179620971 on Build GRCh38
rs781472878 Chr.5: 179621066-179621076 on Build GRCh38
rs200897727 Chr.5: 179620880-179620880 on Build GRCh38
rs183132589 Chr.5: 179620799-179620799 on Build GRCh38
rs755523819 Chr.5: 179621141-179621141 on Build GRCh38
rs529901479 Chr.5: 179621114-179621114 on Build GRCh38
rs868803469 Chr.5: 179620963-179620963 on Build GRCh38
rs911453147 Chr.5: 179621065-179621065 on Build GRCh38
rs901576992 Chr.5: 179620776-179620776 on Build GRCh38
rs768578841 Chr.5: 179620875-179620875 on Build GRCh38
rs557426153 Chr.5: 179620893-179620893 on Build GRCh38
rs757230778 Chr.5: 179620975-179620983 on Build GRCh38
rs543359018 Chr.5: 179620730-179620730 on Build GRCh38
rs1045724064 Chr.5: 179620778-179620778 on Build GRCh38
rs1036196550 Chr.5: 179621100-179621100 on Build GRCh38
rs77085994 Chr.5: 179620878-179620878 on Build GRCh38
rs569166833 Chr.5: 179621090-179621090 on Build GRCh38
rs767934872 Chr.5: 179620923-179620923 on Build GRCh38
rs771792452 Chr.5: 179620842-179620842 on Build GRCh38
rs149952607 Chr.5: 179621018-179621018 on Build GRCh38
rs772752602 Chr.5: 179621074-179621074 on Build GRCh38
rs777866069 Chr.5: 179620862-179620863 on Build GRCh38
rs532542581 Chr.5: 179620840-179620840 on Build GRCh38
rs779008575 Chr.5: 179621042-179621042 on Build GRCh38
rs746904158 Chr.5: 179620876-179620876 on Build GRCh38
rs1048898797 Chr.5: 179620690-179620690 on Build GRCh38
rs766834461 Chr.5: 179620882-179620882 on Build GRCh38
rs575969318 Chr.5: 179620686-179620686 on Build GRCh38
rs375727277 Chr.5: 179621066-179621066 on Build GRCh38
rs890085556 Chr.5: 179620746-179620746 on Build GRCh38
rs921686628 Chr.5: 179620674-179620674 on Build GRCh38
rs865913754 Chr.5: 179621139-179621139 on Build GRCh38
rs759984048 Chr.5: 179620896-179620896 on Build GRCh38
rs931687497 Chr.5: 179620684-179620684 on Build GRCh38
rs917843565 Chr.5: 179621116-179621116 on Build GRCh38
rs757321283 Chr.5: 179621039-179621039 on Build GRCh38
rs768627628 Chr.5: 179621058-179621058 on Build GRCh38
rs865868784 Chr.5: 179620871-179620871 on Build GRCh38

View Full Product Details