Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31688663-31689214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD16A
Entrez Gene ID
7920
Gene Name
abhydrolase domain containing 16A
Gene Aliases
BAT5, D6S82E, NG26, PP199
UniGene
Hs.388188
Gene Chromosome Location
Chr.6: 31686949-31703360 on Build GRCh38
Transcripts
NM_001177515.1 NM_021160.2 NR_033488.1 NR_033489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391773 c.1193T>A p.L398Q Chr.6: 31688780-31688780 on Build GRCh38
5799616 c.1108G>A p.D370N Chr.6: 31689093-31689093 on Build GRCh38
296274 c.1174C>T p.P392S Chr.6: 31689027-31689027 on Build GRCh38
4971541 c.1190G>A p.G397D Chr.6: 31688783-31688783 on Build GRCh38
7000216 c.1250+4A>G p.? Chr.6: 31688719-31688719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773450400 Chr.6: 31688790-31688790 on Build GRCh38
rs757211413 Chr.6: 31689143-31689143 on Build GRCh38
rs147681590 Chr.6: 31689115-31689115 on Build GRCh38
rs946278131 Chr.6: 31688859-31688859 on Build GRCh38
rs779683322 Chr.6: 31689054-31689054 on Build GRCh38
rs761135642 Chr.6: 31689116-31689116 on Build GRCh38
rs988587880 Chr.6: 31688819-31688819 on Build GRCh38
rs906784117 Chr.6: 31688773-31688773 on Build GRCh38
rs73398298 Chr.6: 31688892-31688892 on Build GRCh38
rs992490716 Chr.6: 31688707-31688707 on Build GRCh38
rs765745368 Chr.6: 31688902-31688902 on Build GRCh38
rs375396631 Chr.6: 31689071-31689071 on Build GRCh38
rs760870939 Chr.6: 31688824-31688824 on Build GRCh38
rs750632865 Chr.6: 31688703-31688703 on Build GRCh38
rs762085405 Chr.6: 31688700-31688700 on Build GRCh38
rs755509459 Chr.6: 31689141-31689142 on Build GRCh38
rs537802295 Chr.6: 31688738-31688738 on Build GRCh38
rs1039409651 Chr.6: 31688917-31688917 on Build GRCh38
rs373259819 Chr.6: 31689005-31689005 on Build GRCh38
rs1052605303 Chr.6: 31689148-31689148 on Build GRCh38
rs1045029764 Chr.6: 31689091-31689091 on Build GRCh38
rs1003694889 Chr.6: 31688776-31688776 on Build GRCh38
rs766555812 Chr.6: 31688829-31688829 on Build GRCh38
rs763493393 Chr.6: 31688686-31688686 on Build GRCh38
rs180999181 Chr.6: 31688690-31688690 on Build GRCh38
rs748642978 Chr.6: 31689097-31689097 on Build GRCh38
rs745533005 Chr.6: 31689033-31689033 on Build GRCh38
rs375029790 Chr.6: 31688770-31688770 on Build GRCh38
rs534868195 Chr.6: 31688867-31688867 on Build GRCh38
rs75509151 Chr.6: 31688737-31688737 on Build GRCh38
rs559219579 Chr.6: 31688783-31688783 on Build GRCh38
rs1025278543 Chr.6: 31688693-31688693 on Build GRCh38
rs774899370 Chr.6: 31689073-31689073 on Build GRCh38
rs45480896 Chr.6: 31688858-31688858 on Build GRCh38
rs780892882 Chr.6: 31689030-31689030 on Build GRCh38
rs898278382 Chr.6: 31688950-31688950 on Build GRCh38
rs891287001 Chr.6: 31688888-31688888 on Build GRCh38
rs12193561 Chr.6: 31688751-31688751 on Build GRCh38
rs962348613 Chr.6: 31688683-31688683 on Build GRCh38
rs772479646 Chr.6: 31689108-31689108 on Build GRCh38
rs542752606 Chr.6: 31689136-31689136 on Build GRCh38
rs886783463 Chr.6: 31689163-31689163 on Build GRCh38
rs755670742 Chr.6: 31688998-31688999 on Build GRCh38
rs767734658 Chr.6: 31688702-31688702 on Build GRCh38
rs566154591 Chr.6: 31688698-31688698 on Build GRCh38
rs764084316 Chr.6: 31688971-31688971 on Build GRCh38
rs757057541 Chr.6: 31689007-31689007 on Build GRCh38
rs751486344 Chr.6: 31689142-31689142 on Build GRCh38
rs368223395 Chr.6: 31689034-31689034 on Build GRCh38
rs532361118 Chr.6: 31688685-31688685 on Build GRCh38
rs777024322 Chr.6: 31689131-31689131 on Build GRCh38
rs147830631 Chr.6: 31688740-31688740 on Build GRCh38
rs554220316 Chr.6: 31689155-31689155 on Build GRCh38
rs929763896 Chr.6: 31688920-31688920 on Build GRCh38
rs754321360 Chr.6: 31689016-31689016 on Build GRCh38
rs35552493 Chr.6: 31689015-31689016 on Build GRCh38
rs570037754 Chr.6: 31688767-31688767 on Build GRCh38
rs145460711 Chr.6: 31689050-31689050 on Build GRCh38
rs764427188 Chr.6: 31688687-31688687 on Build GRCh38
rs767820748 Chr.6: 31688784-31688784 on Build GRCh38
rs775756995 Chr.6: 31688739-31688739 on Build GRCh38
rs745559290 Chr.6: 31688750-31688750 on Build GRCh38
rs751478666 Chr.6: 31688979-31688979 on Build GRCh38
rs759844087 Chr.6: 31688713-31688713 on Build GRCh38
rs948111810 Chr.6: 31689064-31689064 on Build GRCh38
rs748753051 Chr.6: 31689055-31689055 on Build GRCh38
rs752679116 Chr.6: 31688714-31688714 on Build GRCh38
rs565029243 Chr.6: 31688682-31688682 on Build GRCh38
rs759644353 Chr.6: 31689134-31689134 on Build GRCh38
rs570683409 Chr.6: 31688837-31688837 on Build GRCh38
rs758469978 Chr.6: 31688723-31688723 on Build GRCh38
rs756195769 Chr.6: 31688706-31688706 on Build GRCh38
rs769265532 Chr.6: 31688766-31688766 on Build GRCh38
rs976643307 Chr.6: 31688914-31688914 on Build GRCh38
rs755114430 Chr.6: 31688838-31688838 on Build GRCh38
rs756833134 Chr.6: 31689041-31689041 on Build GRCh38
rs1048475346 Chr.6: 31688974-31688974 on Build GRCh38
rs35905334 Chr.6: 31688783-31688783 on Build GRCh38
rs766643400 Chr.6: 31689122-31689122 on Build GRCh38
rs1021756077 Chr.6: 31688826-31688826 on Build GRCh38
rs753332134 Chr.6: 31688916-31688916 on Build GRCh38
rs139631157 Chr.6: 31688782-31688782 on Build GRCh38
rs138255252 Chr.6: 31688789-31688789 on Build GRCh38

View Full Product Details