Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39744615-39745013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM2
Entrez Gene ID
2515
Gene Name
ADAM metallopeptidase domain 2
Gene Aliases
CRYN1, CRYN2, CT15, FTNB, PH-30b, PH30, PH30-beta
UniGene
Hs.177959
Gene Chromosome Location
Chr.8: 39743735-39838289 on Build GRCh38
Transcripts
NM_001278113.1 NM_001278114.1 NM_001464.4 XM_005273468.1 XM_011544479.1 XM_011544480.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
750402 c.2197C>G p.P733A Chr.8: 39744871-39744871 on Build GRCh38
118985 c.2175-2A>T p.? Chr.8: 39744895-39744895 on Build GRCh38
5409569 c.2203G>A p.G735R Chr.8: 39744865-39744865 on Build GRCh38
6442295 c.2180C>T p.P727L Chr.8: 39744888-39744888 on Build GRCh38
5760661 c.2175-3C>T p.? Chr.8: 39744896-39744896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955508025 Chr.8: 39744925-39744925 on Build GRCh38
rs753405132 Chr.8: 39744879-39744879 on Build GRCh38
rs961646007 Chr.8: 39744949-39744949 on Build GRCh38
rs563152142 Chr.8: 39744803-39744803 on Build GRCh38
rs905263632 Chr.8: 39744707-39744707 on Build GRCh38
rs572046273 Chr.8: 39744716-39744716 on Build GRCh38
rs768870022 Chr.8: 39744943-39744943 on Build GRCh38
rs139921527 Chr.8: 39744747-39744747 on Build GRCh38
rs767373655 Chr.8: 39744868-39744868 on Build GRCh38
rs757023219 Chr.8: 39744890-39744890 on Build GRCh38
rs902854812 Chr.8: 39744754-39744754 on Build GRCh38
rs938079557 Chr.8: 39744948-39744948 on Build GRCh38
rs961638908 Chr.8: 39744691-39744691 on Build GRCh38
rs144646998 Chr.8: 39744870-39744870 on Build GRCh38
rs546128626 Chr.8: 39744852-39744852 on Build GRCh38
rs919413830 Chr.8: 39744655-39744655 on Build GRCh38
rs1055665687 Chr.8: 39744722-39744722 on Build GRCh38
rs909559508 Chr.8: 39744644-39744644 on Build GRCh38
rs548292585 Chr.8: 39744922-39744922 on Build GRCh38
rs763238199 Chr.8: 39744924-39744925 on Build GRCh38
rs747273426 Chr.8: 39744829-39744829 on Build GRCh38
rs780369622 Chr.8: 39744917-39744917 on Build GRCh38
rs759462015 Chr.8: 39744864-39744864 on Build GRCh38
rs868494823 Chr.8: 39744937-39744937 on Build GRCh38
rs769080149 Chr.8: 39744834-39744834 on Build GRCh38
rs113905354 Chr.8: 39744787-39744787 on Build GRCh38
rs373986909 Chr.8: 39744962-39744962 on Build GRCh38
rs866076726 Chr.8: 39744710-39744710 on Build GRCh38
rs961108210 Chr.8: 39744865-39744865 on Build GRCh38
rs763983947 Chr.8: 39744878-39744878 on Build GRCh38
rs972385111 Chr.8: 39744652-39744652 on Build GRCh38
rs571014286 Chr.8: 39744685-39744685 on Build GRCh38
rs190593179 Chr.8: 39744779-39744779 on Build GRCh38
rs953744785 Chr.8: 39744808-39744808 on Build GRCh38
rs534306958 Chr.8: 39744718-39744718 on Build GRCh38
rs997988299 Chr.8: 39744772-39744772 on Build GRCh38
rs187467931 Chr.8: 39744743-39744743 on Build GRCh38
rs112011537 Chr.8: 39744776-39744776 on Build GRCh38
rs369846105 Chr.8: 39744791-39744791 on Build GRCh38
rs949547164 Chr.8: 39744679-39744679 on Build GRCh38
rs748253345 Chr.8: 39744742-39744742 on Build GRCh38
rs377567199 Chr.8: 39744900-39744900 on Build GRCh38
rs1016609655 Chr.8: 39744863-39744863 on Build GRCh38
rs780561437 Chr.8: 39744828-39744828 on Build GRCh38
rs374318513 Chr.8: 39744897-39744897 on Build GRCh38
rs908561066 Chr.8: 39744673-39744673 on Build GRCh38
rs773334201 Chr.8: 39744862-39744862 on Build GRCh38
rs757987510 Chr.8: 39744910-39744910 on Build GRCh38
rs762135426 Chr.8: 39744839-39744839 on Build GRCh38
rs942260708 Chr.8: 39744646-39744646 on Build GRCh38
rs1053836154 Chr.8: 39744985-39744985 on Build GRCh38
rs901092221 Chr.8: 39744761-39744761 on Build GRCh38
rs949688462 Chr.8: 39744907-39744907 on Build GRCh38
rs553387711 Chr.8: 39744711-39744711 on Build GRCh38
rs148475883 Chr.8: 39744927-39744927 on Build GRCh38
rs1045643278 Chr.8: 39744751-39744751 on Build GRCh38
rs915728874 Chr.8: 39744729-39744729 on Build GRCh38
rs377355710 Chr.8: 39744838-39744838 on Build GRCh38
rs111292703 Chr.8: 39744708-39744708 on Build GRCh38
rs562025559 Chr.8: 39744959-39744959 on Build GRCh38
rs143373254 Chr.8: 39744809-39744809 on Build GRCh38

View Full Product Details