Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRAT
Entrez Gene ID
9227
Gene Name
lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
Gene Aliases
LCA14
Chromosome Location
Chr.4: 154740841 - 154753119 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004744.4 NP_004735.2 1 - 2 252 75
GenBank mRNA AF071510.1 - 1 - 2 91 75
AK292598.1 - 1 - 2 252 75
AY546086.1 - 1 - 2 223 75

View Full Product Details