The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143820.1 | NP_001137292.1 | 9 - 10 | 1326 | 99 |
NM_001162422.1 | NP_001155894.1 | 3 - 4 | 777 | 99 | |
NM_005238.3 | NP_005229.1 | 7 - 8 | 1425 | 99 | |
XM_011542649.2 | XP_011540951.1 | 10 - 11 | 1462 | 99 | |
XM_011542650.1 | XP_011540952.1 | 8 - 9 | 1917 | 99 | |
XM_011542651.1 | XP_011540953.1 | 8 - 9 | 1096 | 99 | |
XM_011542652.1 | XP_011540954.1 | 6 - 7 | 1554 | 99 | |
XM_017017314.1 | XP_016872803.1 | 9 - 10 | 1315 | 99 | |
XM_017017315.1 | XP_016872804.1 | 8 - 9 | 1241 | 99 | |
XM_017017316.1 | XP_016872805.1 | 8 - 9 | 17432 | 99 | |
XM_017017317.1 | XP_016872806.1 | 8 - 9 | 1100 | 99 | |
GenBank mRNA | AK291840.1 | - | 7 - 8 | 1419 | 99 |
AK301405.1 | - | 8 - 9 | 1348 | 99 | |
AY943926.1 | - | 3 - 4 | 461 | 99 | |
BT019452.1 | - | 7 - 8 | 1109 | 99 | |
BT019453.1 | - | 7 - 8 | 1109 | 99 | |
BX640634.1 | - | 9 - 10 | 1326 | 99 | |
CR542254.1 | - | 7 - 8 | 1109 | 99 | |
J04101.1 | - | 7 - 8 | 1228 | 99 | |
X14798.1 | - | 7 - 8 | 1387 | 99 |