The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012089.2 | NP_036221.2 | 4 - 5 | 1095 | 105 |
XM_011544135.2 | XP_011542437.1 | 4 - 5 | 1866 | 105 | |
XM_011544136.2 | XP_011542438.1 | 2 - 3 | 332 | 105 | |
GenBank mRNA | AB013380.1 | - | 4 - 5 | 1085 | 105 |
AF216833.1 | - | 4 - 5 | 1095 | 105 | |
AK172794.1 | - | 3 - 4 | 351 | 105 | |
BC064930.1 | - | 4 - 5 | 1090 | 105 |