Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7066670-7067258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1S
Entrez Gene ID
716
Gene Name
complement component 1, s subcomponent
Gene Aliases
-
UniGene
Hs.458355
Gene Chromosome Location
Chr.12: 7060676-7071032 on Build GRCh38
Transcripts
NM_001734.3 NM_201442.2 XM_005253760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1735248 c.1023T>G p.C341W Chr.12: 7067074-7067074 on Build GRCh38
4667258 c.992G>A p.R331H Chr.12: 7067043-7067043 on Build GRCh38
3872441 c.1049C>T p.S350F Chr.12: 7067100-7067100 on Build GRCh38
1676693 c.1063C>T p.Q355* Chr.12: 7067114-7067114 on Build GRCh38
943219 c.1017G>A p.S339S Chr.12: 7067068-7067068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372578611 Chr.12: 7067179-7067179 on Build GRCh38
rs781956061 Chr.12: 7067054-7067054 on Build GRCh38
rs192346248 Chr.12: 7067097-7067097 on Build GRCh38
rs182893405 Chr.12: 7066781-7066781 on Build GRCh38
rs782146838 Chr.12: 7066712-7066712 on Build GRCh38
rs782641838 Chr.12: 7067185-7067185 on Build GRCh38
rs782288924 Chr.12: 7067056-7067056 on Build GRCh38
rs1025819470 Chr.12: 7066858-7066858 on Build GRCh38
rs781913781 Chr.12: 7067067-7067067 on Build GRCh38
rs781915882 Chr.12: 7066787-7066787 on Build GRCh38
rs112225079 Chr.12: 7067118-7067118 on Build GRCh38
rs75589246 Chr.12: 7066909-7066909 on Build GRCh38
rs187530187 Chr.12: 7066962-7066962 on Build GRCh38
rs782551491 Chr.12: 7066724-7066724 on Build GRCh38
rs782254018 Chr.12: 7066946-7066946 on Build GRCh38
rs150742484 Chr.12: 7067152-7067152 on Build GRCh38
rs782408925 Chr.12: 7067040-7067040 on Build GRCh38
rs782183288 Chr.12: 7067035-7067035 on Build GRCh38
rs782497025 Chr.12: 7066748-7066748 on Build GRCh38
rs1046250658 Chr.12: 7066730-7066730 on Build GRCh38
rs782810160 Chr.12: 7067198-7067198 on Build GRCh38
rs142937694 Chr.12: 7066898-7066898 on Build GRCh38
rs147394609 Chr.12: 7067127-7067127 on Build GRCh38
rs782700707 Chr.12: 7067084-7067084 on Build GRCh38
rs911656234 Chr.12: 7066872-7066872 on Build GRCh38
rs965813370 Chr.12: 7066873-7066873 on Build GRCh38
rs782328086 Chr.12: 7067166-7067166 on Build GRCh38
rs782214906 Chr.12: 7067146-7067146 on Build GRCh38
rs782523614 Chr.12: 7066994-7066994 on Build GRCh38
rs782620950 Chr.12: 7067141-7067141 on Build GRCh38
rs781854420 Chr.12: 7066735-7066735 on Build GRCh38
rs782223222 Chr.12: 7067007-7067007 on Build GRCh38
rs782666102 Chr.12: 7067117-7067117 on Build GRCh38
rs561586059 Chr.12: 7067098-7067098 on Build GRCh38
rs782449714 Chr.12: 7067115-7067115 on Build GRCh38
rs782773916 Chr.12: 7067077-7067077 on Build GRCh38
rs782584563 Chr.12: 7067031-7067031 on Build GRCh38
rs913129197 Chr.12: 7066932-7066932 on Build GRCh38
rs782362588 Chr.12: 7067043-7067043 on Build GRCh38
rs782241051 Chr.12: 7067227-7067227 on Build GRCh38
rs962971067 Chr.12: 7066770-7066770 on Build GRCh38
rs782257320 Chr.12: 7066758-7066758 on Build GRCh38
rs782336370 Chr.12: 7067018-7067018 on Build GRCh38
rs951563926 Chr.12: 7066859-7066859 on Build GRCh38
rs995718954 Chr.12: 7066791-7066791 on Build GRCh38
rs782535131 Chr.12: 7067068-7067068 on Build GRCh38
rs986965980 Chr.12: 7066861-7066861 on Build GRCh38
rs782377050 Chr.12: 7067136-7067136 on Build GRCh38
rs782476821 Chr.12: 7067089-7067089 on Build GRCh38
rs907266016 Chr.12: 7066731-7066731 on Build GRCh38
rs782070807 Chr.12: 7067058-7067058 on Build GRCh38
rs34569978 Chr.12: 7067187-7067188 on Build GRCh38
rs782671818 Chr.12: 7066921-7066921 on Build GRCh38
rs782655255 Chr.12: 7066809-7066809 on Build GRCh38
rs782621962 Chr.12: 7066995-7066995 on Build GRCh38
rs782648242 Chr.12: 7066955-7066955 on Build GRCh38
rs974852182 Chr.12: 7066887-7066887 on Build GRCh38
rs782040678 Chr.12: 7066928-7066928 on Build GRCh38

View Full Product Details