Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134260878-134261445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD8
Entrez Gene ID
27034
Gene Name
acyl-CoA dehydrogenase family member 8
Gene Aliases
ACAD-8, ARC42
UniGene
Hs.14791
Gene Chromosome Location
Chr.11: 134253532-134265858 on Build GRCh38
Transcripts
NM_014384.2 XM_005271505.3 XM_011542750.2 XM_017017542.1 XM_017017543.1 XM_017017544.1 XM_017017545.1 XM_017017546.1 XM_017017547.1 XM_017017548.1 XM_017017549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5212423 c.632T>C p.L211P Chr.11: 134261359-134261359 on Build GRCh38
4019543 c.904C>T p.R302W Chr.11: 134261337-134261337 on Build GRCh38
1475279 c.926T>C p.L309P Chr.11: 134261359-134261359 on Build GRCh38
6265457 c.738G>T p.V246V Chr.11: 134261076-134261076 on Build GRCh38
5838209 c.674G>A p.R225Q Chr.11: 134261338-134261338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374459415 Chr.11: 134261082-134261082 on Build GRCh38
rs745524886 Chr.11: 134261371-134261371 on Build GRCh38
rs562769668 Chr.11: 134261220-134261220 on Build GRCh38
rs376839376 Chr.11: 134261021-134261021 on Build GRCh38
rs371156848 Chr.11: 134261160-134261160 on Build GRCh38
rs760641912 Chr.11: 134261293-134261293 on Build GRCh38
rs756980031 Chr.11: 134261357-134261357 on Build GRCh38
rs1024578942 Chr.11: 134261233-134261233 on Build GRCh38
rs376919619 Chr.11: 134261039-134261039 on Build GRCh38
rs75844584 Chr.11: 134261023-134261023 on Build GRCh38
rs757386559 Chr.11: 134261065-134261065 on Build GRCh38
rs77067193 Chr.11: 134261217-134261217 on Build GRCh38
rs1033152085 Chr.11: 134261146-134261146 on Build GRCh38
rs74971988 Chr.11: 134260914-134260914 on Build GRCh38
rs770612690 Chr.11: 134261194-134261194 on Build GRCh38
rs374265813 Chr.11: 134261161-134261161 on Build GRCh38
rs201593770 Chr.11: 134261097-134261097 on Build GRCh38
rs533310201 Chr.11: 134260958-134260958 on Build GRCh38
rs754203991 Chr.11: 134261348-134261348 on Build GRCh38
rs774494949 Chr.11: 134261402-134261402 on Build GRCh38
rs775352104 Chr.11: 134261288-134261288 on Build GRCh38
rs767383153 Chr.11: 134261133-134261133 on Build GRCh38
rs746960051 Chr.11: 134261068-134261068 on Build GRCh38
rs766230652 Chr.11: 134261351-134261353 on Build GRCh38
rs183752972 Chr.11: 134261076-134261076 on Build GRCh38
rs142790411 Chr.11: 134261144-134261144 on Build GRCh38
rs200754189 Chr.11: 134261105-134261105 on Build GRCh38
rs146070958 Chr.11: 134261100-134261100 on Build GRCh38
rs930743838 Chr.11: 134261241-134261241 on Build GRCh38
rs200330956 Chr.11: 134261279-134261279 on Build GRCh38
rs759781523 Chr.11: 134261408-134261408 on Build GRCh38
rs775062752 Chr.11: 134261320-134261320 on Build GRCh38
rs967555931 Chr.11: 134260960-134260960 on Build GRCh38
rs774487413 Chr.11: 134261121-134261121 on Build GRCh38
rs36115758 Chr.11: 134261139-134261139 on Build GRCh38
rs200780328 Chr.11: 134261206-134261206 on Build GRCh38
rs77052364 Chr.11: 134260993-134260993 on Build GRCh38
rs967341745 Chr.11: 134261246-134261246 on Build GRCh38
rs950055547 Chr.11: 134261256-134261256 on Build GRCh38
rs974026249 Chr.11: 134261421-134261421 on Build GRCh38
rs775598743 Chr.11: 134261417-134261417 on Build GRCh38
rs750643088 Chr.11: 134261152-134261152 on Build GRCh38
rs537831211 Chr.11: 134261096-134261096 on Build GRCh38
rs753192386 Chr.11: 134261328-134261328 on Build GRCh38
rs908368010 Chr.11: 134261049-134261049 on Build GRCh38
rs763133668 Chr.11: 134261218-134261218 on Build GRCh38
rs747871623 Chr.11: 134261387-134261387 on Build GRCh38
rs754886392 Chr.11: 134261043-134261043 on Build GRCh38
rs778257880 Chr.11: 134261260-134261260 on Build GRCh38
rs961266766 Chr.11: 134261419-134261419 on Build GRCh38
rs777635296 Chr.11: 134261188-134261188 on Build GRCh38
rs199738913 Chr.11: 134261185-134261185 on Build GRCh38
rs74891173 Chr.11: 134260976-134260976 on Build GRCh38
rs1014502442 Chr.11: 134261124-134261124 on Build GRCh38
rs973890269 Chr.11: 134261214-134261214 on Build GRCh38
rs779587719 Chr.11: 134261377-134261377 on Build GRCh38
rs369676427 Chr.11: 134261423-134261423 on Build GRCh38
rs977568732 Chr.11: 134260964-134260964 on Build GRCh38
rs578035325 Chr.11: 134261229-134261229 on Build GRCh38
rs754767707 Chr.11: 134261352-134261352 on Build GRCh38
rs201188279 Chr.11: 134261267-134261267 on Build GRCh38
rs768533799 Chr.11: 134261378-134261378 on Build GRCh38
rs367586207 Chr.11: 134261234-134261234 on Build GRCh38
rs899220061 Chr.11: 134261101-134261101 on Build GRCh38
rs762569504 Chr.11: 134261302-134261302 on Build GRCh38
rs578153873 Chr.11: 134261257-134261257 on Build GRCh38
rs144566960 Chr.11: 134261301-134261301 on Build GRCh38
rs1028456640 Chr.11: 134261343-134261343 on Build GRCh38
rs121908421 Chr.11: 134261300-134261300 on Build GRCh38
rs543302312 Chr.11: 134261413-134261413 on Build GRCh38
rs1034778172 Chr.11: 134260910-134260910 on Build GRCh38
rs998953069 Chr.11: 134261145-134261145 on Build GRCh38
rs986266918 Chr.11: 134261268-134261268 on Build GRCh38
rs748933800 Chr.11: 134261189-134261189 on Build GRCh38
rs995360618 Chr.11: 134261109-134261109 on Build GRCh38
rs986435769 Chr.11: 134261384-134261384 on Build GRCh38
rs768482407 Chr.11: 134261295-134261295 on Build GRCh38
rs757519037 Chr.11: 134261142-134261142 on Build GRCh38
rs781637075 Chr.11: 134261177-134261177 on Build GRCh38
rs763007237 Chr.11: 134261025-134261025 on Build GRCh38
rs121908422 Chr.11: 134261338-134261338 on Build GRCh38
rs758914454 Chr.11: 134261069-134261069 on Build GRCh38
rs776832750 Chr.11: 134261087-134261087 on Build GRCh38
rs77368661 Chr.11: 134261186-134261186 on Build GRCh38
rs770663870 Chr.11: 134261278-134261278 on Build GRCh38
rs759600553 Chr.11: 134261319-134261319 on Build GRCh38
rs1027985109 Chr.11: 134261178-134261178 on Build GRCh38
rs575969865 Chr.11: 134261209-134261209 on Build GRCh38
rs372030555 Chr.11: 134261213-134261213 on Build GRCh38
rs370199799 Chr.11: 134261053-134261053 on Build GRCh38
rs572170782 Chr.11: 134261163-134261163 on Build GRCh38
rs1040014399 Chr.11: 134260935-134260935 on Build GRCh38
rs1014946159 Chr.11: 134260939-134260939 on Build GRCh38
rs766531579 Chr.11: 134261307-134261307 on Build GRCh38
rs776405341 Chr.11: 134260997-134260997 on Build GRCh38
rs750026561 Chr.11: 134261030-134261031 on Build GRCh38
rs957134555 Chr.11: 134260977-134260977 on Build GRCh38
rs757899755 Chr.11: 134261373-134261373 on Build GRCh38
rs761351710 Chr.11: 134261009-134261009 on Build GRCh38
rs368653287 Chr.11: 134261122-134261122 on Build GRCh38
rs765982422 Chr.11: 134261113-134261113 on Build GRCh38
rs780609527 Chr.11: 134261072-134261072 on Build GRCh38
rs781486443 Chr.11: 134261253-134261253 on Build GRCh38
rs376407410 Chr.11: 134261395-134261395 on Build GRCh38
rs755402140 Chr.11: 134261176-134261176 on Build GRCh38
rs756612574 Chr.11: 134261337-134261337 on Build GRCh38
rs372904463 Chr.11: 134261383-134261383 on Build GRCh38
rs773964375 Chr.11: 134261197-134261197 on Build GRCh38
rs767201554 Chr.11: 134261051-134261051 on Build GRCh38
rs372337818 Chr.11: 134261202-134261202 on Build GRCh38
rs372958578 Chr.11: 134261032-134261032 on Build GRCh38
rs771611393 Chr.11: 134261286-134261286 on Build GRCh38
rs201716113 Chr.11: 134261254-134261254 on Build GRCh38
rs778637892 Chr.11: 134261365-134261365 on Build GRCh38
rs527890266 Chr.11: 134261315-134261315 on Build GRCh38
rs1017822780 Chr.11: 134261297-134261297 on Build GRCh38
rs766360602 Chr.11: 134261035-134261035 on Build GRCh38

View Full Product Details