Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NBPF9
- Entrez Gene ID
-
400818
- Gene Name
- neuroblastoma breakpoint family member 9
- Gene Aliases
-
AE01
- Chromosome Location
-
Chr.1: 149054033 - 149090861 on Build GRCh38
- Gene Symbol
- LOC102724250
- Entrez Gene ID
-
102724250
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1|NT_187366: 4611 - 41174 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_011546188.2 |
XP_011544490.1 |
20 - 21 |
11393 |
189 |
- Gene Symbol
- NBPF1
- Entrez Gene ID
-
55672
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
-
AB13, AB14, AB23, AD2, NBG, NBPF
- Chromosome Location
-
Chr.1: 16562427 - 16613605 on Build GRCh38
- Gene Symbol
- NBPF10
- Entrez Gene ID
-
100132406
- Gene Name
- neuroblastoma breakpoint family member 10
- Gene Aliases
-
AB6, AG1, NBPF9
- Chromosome Location
-
Chr.1: 146066318 - 146144804 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF379616.1 |
- |
7 - 8 |
1059 |
189 |
AF380580.1 |
- |
13 - 14 |
1498 |
186 |
AY894565.1 |
- |
17 - 18 |
2324 |
186 |
BX538005.1 |
- |
26 - 27 |
3759 |
186 |
- Gene Symbol
- NBPF12
- Entrez Gene ID
-
149013
- Gene Name
- neuroblastoma breakpoint family member 12
- Gene Aliases
-
COAS1, KIAA1245
- Chromosome Location
-
Chr.1: 146938545 - 146996093 on Build GRCh38
- Gene Symbol
- NBPF14
- Entrez Gene ID
-
25832
- Gene Name
- neuroblastoma breakpoint family member 14
- Gene Aliases
-
DJ328E19.C1.1, NBPF
- Chromosome Location
-
Chr.1: 148531385 - 148607636 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF379626.1 |
- |
10 - 11 |
1155 |
189 |
AL117237.1 |
- |
20 - 21 |
2431 |
186 |
- Gene Symbol
- NBPF15
- Entrez Gene ID
-
284565
- Gene Name
- neuroblastoma breakpoint family member 15
- Gene Aliases
-
AB14, AG3, NBPF16
- Chromosome Location
-
Chr.1: 144421386 - 144459464 on Build GRCh38
- Gene Symbol
- NBPF19
- Entrez Gene ID
-
101060226
- Gene Name
- neuroblastoma breakpoint family member 19
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 149472955 - 149556256 on Build GRCh38
- Gene Symbol
- NBPF20
- Entrez Gene ID
-
100288142
- Gene Name
- neuroblastoma breakpoint family member 20
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 145289900 - 145405778 on Build GRCh38
- Gene Symbol
- NBPF25P
- Entrez Gene ID
-
101929780
- Gene Name
- neuroblastoma breakpoint family member 25, pseudogene
- Gene Aliases
-
WI2-925H4.1
- Chromosome Location
-
Chr.1: 145572345 - 145607858 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104217.1 |
|
17 - 18 |
2121 |
189 |
GenBank mRNA |
AK125792.1 |
- |
17 - 18 |
2121 |
189 |
- Gene Symbol
- NBPF26
- Entrez Gene ID
-
101060684
- Gene Name
- neuroblastoma breakpoint family member 26
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 120805561 - 120842215 on Build GRCh38
- Gene Symbol
- NBPF8
- Entrez Gene ID
-
728841
- Gene Name
- neuroblastoma breakpoint family member 8
- Gene Aliases
-
NBPF8P
- Chromosome Location
-
Chr.1: 120436351 - 120467844 on Build GRCh38
View Full Product Details