Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 65225895-65226477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK4
Entrez Gene ID
205
Gene Name
adenylate kinase 4
Gene Aliases
AK 4, AK3, AK3L1, AK3L2
UniGene
Hs.10862
Gene Chromosome Location
Chr.1: 65147342-65232145 on Build GRCh38
Transcripts
NM_001005353.2 NM_013410.3 NM_203464.2 XM_017000612.1 XM_017000613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419389 c.560G>A p.S187N Chr.1: 65226065-65226065 on Build GRCh38
6997018 c.585C>T p.S195S Chr.1: 65226090-65226090 on Build GRCh38
5916974 c.592G>A p.E198K Chr.1: 65226097-65226097 on Build GRCh38
7147390 c.563G>A p.R188Q Chr.1: 65226068-65226068 on Build GRCh38
1235262 c.589A>G p.T197A Chr.1: 65226094-65226094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536799600 Chr.1: 65226248-65226248 on Build GRCh38
rs765173381 Chr.1: 65226107-65226107 on Build GRCh38
rs763062182 Chr.1: 65226083-65226083 on Build GRCh38
rs976943311 Chr.1: 65226189-65226189 on Build GRCh38
rs188589000 Chr.1: 65225932-65225932 on Build GRCh38
rs564798376 Chr.1: 65226014-65226014 on Build GRCh38
rs1011941793 Chr.1: 65226038-65226038 on Build GRCh38
rs1021722774 Chr.1: 65225922-65225922 on Build GRCh38
rs749824690 Chr.1: 65226193-65226193 on Build GRCh38
rs746234878 Chr.1: 65226074-65226074 on Build GRCh38
rs1052357734 Chr.1: 65226067-65226067 on Build GRCh38
rs943873223 Chr.1: 65225991-65225991 on Build GRCh38
rs747654942 Chr.1: 65226037-65226037 on Build GRCh38
rs543317459 Chr.1: 65225972-65225972 on Build GRCh38
rs776031990 Chr.1: 65226173-65226173 on Build GRCh38
rs745458985 Chr.1: 65226206-65226208 on Build GRCh38
rs770245271 Chr.1: 65226076-65226076 on Build GRCh38
rs754855850 Chr.1: 65226104-65226104 on Build GRCh38
rs778444672 Chr.1: 65226204-65226205 on Build GRCh38
rs550508905 Chr.1: 65226102-65226102 on Build GRCh38
rs142977116 Chr.1: 65226033-65226033 on Build GRCh38
rs951532891 Chr.1: 65226265-65226265 on Build GRCh38
rs761992146 Chr.1: 65226226-65226226 on Build GRCh38
rs558447583 Chr.1: 65226367-65226367 on Build GRCh38
rs965640817 Chr.1: 65225968-65225968 on Build GRCh38
rs142579030 Chr.1: 65226060-65226060 on Build GRCh38
rs764116417 Chr.1: 65226090-65226090 on Build GRCh38
rs768769636 Chr.1: 65226221-65226221 on Build GRCh38
rs149373720 Chr.1: 65226121-65226121 on Build GRCh38
rs531898415 Chr.1: 65226101-65226101 on Build GRCh38
rs757869386 Chr.1: 65226040-65226040 on Build GRCh38
rs201234545 Chr.1: 65226154-65226154 on Build GRCh38
rs930158726 Chr.1: 65226013-65226013 on Build GRCh38
rs947867749 Chr.1: 65226442-65226442 on Build GRCh38
rs770100726 Chr.1: 65226171-65226171 on Build GRCh38
rs753356079 Chr.1: 65226045-65226045 on Build GRCh38
rs984336609 Chr.1: 65226391-65226391 on Build GRCh38
rs183989159 Chr.1: 65226336-65226336 on Build GRCh38
rs909617258 Chr.1: 65226394-65226395 on Build GRCh38
rs778724734 Chr.1: 65226034-65226034 on Build GRCh38
rs776921061 Chr.1: 65226068-65226068 on Build GRCh38
rs567848952 Chr.1: 65226096-65226096 on Build GRCh38
rs904633741 Chr.1: 65226115-65226115 on Build GRCh38
rs575841967 Chr.1: 65226284-65226284 on Build GRCh38
rs576757716 Chr.1: 65226423-65226423 on Build GRCh38
rs947700107 Chr.1: 65226027-65226027 on Build GRCh38
rs746535040 Chr.1: 65226127-65226127 on Build GRCh38
rs539317041 Chr.1: 65226143-65226143 on Build GRCh38
rs748404457 Chr.1: 65226006-65226006 on Build GRCh38
rs149026313 Chr.1: 65225961-65225961 on Build GRCh38
rs932722204 Chr.1: 65225945-65225945 on Build GRCh38
rs772388249 Chr.1: 65226227-65226227 on Build GRCh38
rs547970998 Chr.1: 65226079-65226079 on Build GRCh38
rs969097506 Chr.1: 65225959-65225959 on Build GRCh38
rs151106010 Chr.1: 65226049-65226049 on Build GRCh38
rs746247285 Chr.1: 65225978-65225978 on Build GRCh38
rs138516264 Chr.1: 65226065-65226065 on Build GRCh38
rs374408153 Chr.1: 65226368-65226368 on Build GRCh38
rs756777375 Chr.1: 65226047-65226047 on Build GRCh38
rs146709398 Chr.1: 65226120-65226120 on Build GRCh38
rs565925334 Chr.1: 65226157-65226157 on Build GRCh38
rs774240631 Chr.1: 65226225-65226225 on Build GRCh38
rs554720574 Chr.1: 65226249-65226249 on Build GRCh38
rs191906974 Chr.1: 65226024-65226024 on Build GRCh38
rs766346242 Chr.1: 65226017-65226017 on Build GRCh38
rs576155198 Chr.1: 65225964-65225964 on Build GRCh38

View Full Product Details