Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
Chromosome Location
Chr.1: 151510859 - 151538692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020770.2 NP_065821.1 17 - 18 3219 64
XM_005245365.4 XP_005245422.1 17 - 18 3231 64
GenBank mRNA AB037740.1 - 17 - 18 3192 64
AF263462.1 - 17 - 18 3226 64
AK290007.1 - 17 - 18 3192 64
BC146657.1 - 16 - 17 3497 64
BC150604.1 - 16 - 17 3098 64
BC152445.1 - 17 - 18 3192 64
BX647343.1 - 2 - 3 1073 64

View Full Product Details