The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166624.1 | NP_001160096.1 | 2 - 3 | 338 | 119 |
NM_021023.5 | NP_066303.2 | 2 - 3 | 338 | 119 | |
GenBank mRNA | AK298459.1 | - | 2 - 3 | 322 | 119 |
BC020687.1 | - | 2 - 3 | 322 | 119 | |
BC058009.1 | - | 2 - 3 | 296 | 119 | |
BC070259.1 | - | 2 - 3 | 325 | 119 | |
X68679.1 | - | 2 - 3 | 274 | 119 |