Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31084562-31085059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4S1
Entrez Gene ID
11154
Gene Name
adaptor related protein complex 4 sigma 1 subunit
Gene Aliases
AP47B, CLA20, CLAPS4, CPSQ6, SPG52
UniGene
Hs.293411
Gene Chromosome Location
Chr.14: 31025106-31096450 on Build GRCh38
Transcripts
NM_001128126.2 NM_001254726.1 NM_001254727.1 NM_001254728.1 NM_001254729.1 NM_007077.4 XM_005267293.4 XM_011536371.2 XM_011536372.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607596 c.372A>G p.P124P Chr.14: 31084774-31084774 on Build GRCh38
4571298 c.426T>C p.F142F Chr.14: 31084828-31084828 on Build GRCh38
4612337 c.467delC p.I158fs*>2 Chr.14: 31084869-31084869 on Build GRCh38
5634486 c.422G>T p.C141F Chr.14: 31084824-31084824 on Build GRCh38
4411848 c.462C>G p.S154S Chr.14: 31084864-31084864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146567471 Chr.14: 31084844-31084844 on Build GRCh38
rs772262664 Chr.14: 31084834-31084834 on Build GRCh38
rs1008412339 Chr.14: 31084632-31084632 on Build GRCh38
rs1049620201 Chr.14: 31084600-31084600 on Build GRCh38
rs747269244 Chr.14: 31084925-31084925 on Build GRCh38
rs749920998 Chr.14: 31084852-31084852 on Build GRCh38
rs774449729 Chr.14: 31084782-31084782 on Build GRCh38
rs370072298 Chr.14: 31084743-31084743 on Build GRCh38
rs769669007 Chr.14: 31084711-31084711 on Build GRCh38
rs200969079 Chr.14: 31084775-31084775 on Build GRCh38
rs896823221 Chr.14: 31084649-31084649 on Build GRCh38
rs754858244 Chr.14: 31084765-31084765 on Build GRCh38
rs528491755 Chr.14: 31084859-31084859 on Build GRCh38
rs760944682 Chr.14: 31084843-31084843 on Build GRCh38
rs776944069 Chr.14: 31084849-31084849 on Build GRCh38
rs551412687 Chr.14: 31084667-31084667 on Build GRCh38
rs372843411 Chr.14: 31084705-31084705 on Build GRCh38
rs776262242 Chr.14: 31084834-31084835 on Build GRCh38
rs797045252 Chr.14: 31084766-31084767 on Build GRCh38
rs945396457 Chr.14: 31084612-31084612 on Build GRCh38
rs374141353 Chr.14: 31084858-31084858 on Build GRCh38
rs112223943 Chr.14: 31084929-31084929 on Build GRCh38
rs367866727 Chr.14: 31085016-31085016 on Build GRCh38
rs987055762 Chr.14: 31084893-31084893 on Build GRCh38
rs562382764 Chr.14: 31084639-31084639 on Build GRCh38
rs864309598 Chr.14: 31084839-31084839 on Build GRCh38
rs760711245 Chr.14: 31084648-31084648 on Build GRCh38
rs864309597 Chr.14: 31084838-31084839 on Build GRCh38
rs775840348 Chr.14: 31084838-31084838 on Build GRCh38
rs750047665 Chr.14: 31084721-31084721 on Build GRCh38
rs778167831 Chr.14: 31084723-31084726 on Build GRCh38
rs1034496187 Chr.14: 31084759-31084759 on Build GRCh38
rs758028160 Chr.14: 31084857-31084857 on Build GRCh38
rs912544646 Chr.14: 31084605-31084605 on Build GRCh38
rs889594850 Chr.14: 31084616-31084616 on Build GRCh38
rs551088859 Chr.14: 31084943-31084943 on Build GRCh38
rs143795469 Chr.14: 31084867-31084867 on Build GRCh38
rs1034528586 Chr.14: 31084951-31084951 on Build GRCh38
rs913347896 Chr.14: 31084766-31084766 on Build GRCh38
rs943852723 Chr.14: 31084887-31084887 on Build GRCh38
rs531600804 Chr.14: 31084967-31084967 on Build GRCh38
rs141134249 Chr.14: 31084961-31084961 on Build GRCh38
rs143728054 Chr.14: 31084879-31084879 on Build GRCh38
rs960193671 Chr.14: 31084762-31084762 on Build GRCh38
rs748006650 Chr.14: 31084847-31084847 on Build GRCh38
rs748041792 Chr.14: 31084864-31084864 on Build GRCh38
rs779204070 Chr.14: 31084759-31084759 on Build GRCh38
rs72670341 Chr.14: 31084932-31084932 on Build GRCh38
rs45530436 Chr.14: 31084903-31084903 on Build GRCh38
rs569507777 Chr.14: 31084956-31084956 on Build GRCh38
rs780982908 Chr.14: 31084772-31084772 on Build GRCh38
rs775509259 Chr.14: 31084923-31084923 on Build GRCh38
rs764797988 Chr.14: 31084851-31084851 on Build GRCh38
rs745936731 Chr.14: 31084897-31084897 on Build GRCh38
rs779794767 Chr.14: 31084739-31084739 on Build GRCh38
rs749329106 Chr.14: 31084758-31084758 on Build GRCh38
rs1041272792 Chr.14: 31084900-31084900 on Build GRCh38
rs768691807 Chr.14: 31084830-31084830 on Build GRCh38
rs766477779 Chr.14: 31084758-31084759 on Build GRCh38
rs1020855024 Chr.14: 31084964-31084964 on Build GRCh38
rs864309596 Chr.14: 31084837-31084837 on Build GRCh38
rs532943159 Chr.14: 31084645-31084645 on Build GRCh38
rs993886872 Chr.14: 31084726-31084726 on Build GRCh38
rs558189943 Chr.14: 31084989-31084989 on Build GRCh38
rs781066963 Chr.14: 31084861-31084861 on Build GRCh38
rs761359721 Chr.14: 31084644-31084644 on Build GRCh38
rs768853634 Chr.14: 31084927-31084927 on Build GRCh38
rs1026588599 Chr.14: 31084735-31084735 on Build GRCh38
rs1001741023 Chr.14: 31084941-31084941 on Build GRCh38
rs376938112 Chr.14: 31084587-31084587 on Build GRCh38
rs745990829 Chr.14: 31084816-31084816 on Build GRCh38
rs1054211482 Chr.14: 31084682-31084682 on Build GRCh38
rs925342190 Chr.14: 31084948-31084948 on Build GRCh38
rs1055161620 Chr.14: 31084958-31084958 on Build GRCh38
rs374600997 Chr.14: 31084774-31084774 on Build GRCh38

View Full Product Details