Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43063185-43063602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA1
Entrez Gene ID
672
Gene Name
BRCA1, DNA repair associated
Gene Aliases
BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53
UniGene
Hs.194143
Gene Chromosome Location
Chr.17: 43044295-43125483 on Build GRCh38
Transcripts
NM_007294.3 NM_007297.3 NM_007298.3 NM_007299.3 NM_007300.3 NR_027676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4861595 c.5216-1G>A p.? Chr.17: 43063374-43063374 on Build GRCh38
6933612 c.5240G>C p.R1747T Chr.17: 43063349-43063349 on Build GRCh38
7343719 c.5191G>C p.E1731Q Chr.17: 43063335-43063335 on Build GRCh38
7163084 c.5218G>A p.V1740M Chr.17: 43063371-43063371 on Build GRCh38
6568488 c.5240_5241delGA p.R1747fs*5 Chr.17: 43063348-43063349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549676175 Chr.17: 43063485-43063485 on Build GRCh38
rs869312520 Chr.17: 43063523-43063523 on Build GRCh38
rs273901752 Chr.17: 43063318-43063330 on Build GRCh38
rs80357104 Chr.17: 43063361-43063361 on Build GRCh38
rs995310435 Chr.17: 43063566-43063566 on Build GRCh38
rs375647048 Chr.17: 43063295-43063295 on Build GRCh38
rs898328356 Chr.17: 43063554-43063554 on Build GRCh38
rs1019506976 Chr.17: 43063483-43063483 on Build GRCh38
rs28897697 Chr.17: 43063369-43063369 on Build GRCh38
rs398122692 Chr.17: 43063340-43063340 on Build GRCh38
rs273901751 Chr.17: 43063331-43063331 on Build GRCh38
rs45471406 Chr.17: 43063386-43063386 on Build GRCh38
rs273901750 Chr.17: 43063320-43063320 on Build GRCh38
rs886040273 Chr.17: 43063370-43063371 on Build GRCh38
rs397509233 Chr.17: 43063364-43063364 on Build GRCh38
rs587781526 Chr.17: 43063370-43063389 on Build GRCh38
rs988193234 Chr.17: 43063431-43063431 on Build GRCh38
rs533222994 Chr.17: 43063532-43063532 on Build GRCh38
rs80357867 Chr.17: 43063350-43063353 on Build GRCh38
rs1025481403 Chr.17: 43063574-43063574 on Build GRCh38
rs397509235 Chr.17: 43063344-43063344 on Build GRCh38
rs483353100 Chr.17: 43063362-43063362 on Build GRCh38
rs886040915 Chr.17: 43063331-43063331 on Build GRCh38
rs565110238 Chr.17: 43063505-43063505 on Build GRCh38
rs561146481 Chr.17: 43063440-43063440 on Build GRCh38
rs1025362882 Chr.17: 43063269-43063269 on Build GRCh38
rs191373374 Chr.17: 43063351-43063351 on Build GRCh38
rs273901746 Chr.17: 43063375-43063375 on Build GRCh38
rs876660702 Chr.17: 43063333-43063333 on Build GRCh38
rs80357171 Chr.17: 43063337-43063337 on Build GRCh38
rs773082120 Chr.17: 43063406-43063406 on Build GRCh38
rs45518438 Chr.17: 43063521-43063521 on Build GRCh38
rs80357730 Chr.17: 43063348-43063349 on Build GRCh38
rs886040274 Chr.17: 43063361-43063365 on Build GRCh38
rs80357291 Chr.17: 43063353-43063353 on Build GRCh38
rs1006793345 Chr.17: 43063209-43063209 on Build GRCh38
rs762444270 Chr.17: 43063506-43063506 on Build GRCh38
rs886040275 Chr.17: 43063362-43063363 on Build GRCh38
rs779136512 Chr.17: 43063210-43063210 on Build GRCh38
rs995674624 Chr.17: 43063235-43063235 on Build GRCh38
rs80357895 Chr.17: 43063369-43063370 on Build GRCh38
rs730882167 Chr.17: 43063359-43063359 on Build GRCh38
rs786202545 Chr.17: 43063375-43063375 on Build GRCh38
rs878853295 Chr.17: 43063344-43063345 on Build GRCh38
rs786203547 Chr.17: 43063349-43063349 on Build GRCh38
rs273901748 Chr.17: 43063439-43063439 on Build GRCh38
rs80357347 Chr.17: 43063347-43063347 on Build GRCh38
rs1018546635 Chr.17: 43063228-43063228 on Build GRCh38
rs370536578 Chr.17: 43063404-43063404 on Build GRCh38
rs80357239 Chr.17: 43063372-43063372 on Build GRCh38
rs80358346 Chr.17: 43063363-43063365 on Build GRCh38
rs765846390 Chr.17: 43063417-43063417 on Build GRCh38
rs80357743 Chr.17: 43063371-43063371 on Build GRCh38
rs80358109 Chr.17: 43063399-43063399 on Build GRCh38
rs56195342 Chr.17: 43063368-43063368 on Build GRCh38
rs774666969 Chr.17: 43063436-43063436 on Build GRCh38
rs878854957 Chr.17: 43063365-43063365 on Build GRCh38
rs397509236 Chr.17: 43063332-43063332 on Build GRCh38
rs80358129 Chr.17: 43063379-43063379 on Build GRCh38
rs375639469 Chr.17: 43063376-43063376 on Build GRCh38
rs34570933 Chr.17: 43063343-43063344 on Build GRCh38
rs41293461 Chr.17: 43063373-43063373 on Build GRCh38
rs753793287 Chr.17: 43063322-43063322 on Build GRCh38
rs8176260 Chr.17: 43063311-43063311 on Build GRCh38
rs876658478 Chr.17: 43063350-43063350 on Build GRCh38
rs749465132 Chr.17: 43063371-43063371 on Build GRCh38
rs776018348 Chr.17: 43063418-43063418 on Build GRCh38
rs1011741395 Chr.17: 43063422-43063422 on Build GRCh38
rs397509234 Chr.17: 43063334-43063347 on Build GRCh38
rs376736915 Chr.17: 43063366-43063366 on Build GRCh38
rs963889320 Chr.17: 43063495-43063495 on Build GRCh38
rs80357975 Chr.17: 43063346-43063349 on Build GRCh38
rs922041187 Chr.17: 43063241-43063241 on Build GRCh38
rs80357501 Chr.17: 43063350-43063350 on Build GRCh38
rs531671458 Chr.17: 43063448-43063448 on Build GRCh38
rs80358004 Chr.17: 43063332-43063332 on Build GRCh38
rs397507244 Chr.17: 43063335-43063335 on Build GRCh38
rs273901749 Chr.17: 43063232-43063232 on Build GRCh38
rs1057517590 Chr.17: 43063370-43063370 on Build GRCh38
rs273901747 Chr.17: 43063415-43063415 on Build GRCh38
rs760559327 Chr.17: 43063413-43063413 on Build GRCh38
rs933399306 Chr.17: 43063462-43063462 on Build GRCh38
rs80358137 Chr.17: 43063374-43063374 on Build GRCh38
rs879254150 Chr.17: 43063354-43063354 on Build GRCh38
rs749668287 Chr.17: 43063437-43063440 on Build GRCh38
rs730881496 Chr.17: 43063340-43063340 on Build GRCh38
rs942629160 Chr.17: 43063516-43063516 on Build GRCh38
rs1007985423 Chr.17: 43063461-43063461 on Build GRCh38

View Full Product Details