Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15353841-15354420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8
Entrez Gene ID
10270
Gene Name
A-kinase anchoring protein 8
Gene Aliases
AKAP 95, AKAP-8, AKAP-95, AKAP95
UniGene
Hs.594496
Gene Chromosome Location
Chr.19: 15353521-15379815 on Build GRCh38
Transcripts
NM_005858.3 XM_011527624.2 XM_017026141.1

SNP Detail

SNP ID Chromosome Location
rs917524332 Chr.19: 15354328-15354328 on Build GRCh38
rs143239866 Chr.19: 15354021-15354022 on Build GRCh38
rs984261778 Chr.19: 15354281-15354281 on Build GRCh38
rs1057168736 Chr.19: 15354100-15354100 on Build GRCh38
rs376982063 Chr.19: 15354101-15354101 on Build GRCh38
rs994607770 Chr.19: 15354229-15354229 on Build GRCh38
rs927507935 Chr.19: 15354069-15354069 on Build GRCh38
rs10413241 Chr.19: 15354109-15354109 on Build GRCh38
rs73508480 Chr.19: 15354138-15354138 on Build GRCh38
rs952846488 Chr.19: 15353978-15353978 on Build GRCh38
rs937623755 Chr.19: 15354087-15354087 on Build GRCh38
rs35132731 Chr.19: 15353930-15353931 on Build GRCh38
rs1019516428 Chr.19: 15353932-15353932 on Build GRCh38
rs145532714 Chr.19: 15354349-15354349 on Build GRCh38
rs542936169 Chr.19: 15354222-15354222 on Build GRCh38
rs1018369444 Chr.19: 15353989-15353989 on Build GRCh38
rs774194328 Chr.19: 15354220-15354220 on Build GRCh38
rs578085706 Chr.19: 15353938-15353938 on Build GRCh38
rs759493984 Chr.19: 15354338-15354338 on Build GRCh38
rs66537228 Chr.19: 15354400-15354400 on Build GRCh38
rs562872953 Chr.19: 15354259-15354259 on Build GRCh38
rs1050238573 Chr.19: 15354139-15354139 on Build GRCh38
rs775262452 Chr.19: 15354127-15354127 on Build GRCh38
rs543323502 Chr.19: 15354375-15354375 on Build GRCh38
rs745314446 Chr.19: 15353960-15353960 on Build GRCh38
rs540428748 Chr.19: 15353979-15353979 on Build GRCh38
rs1019992055 Chr.19: 15354185-15354185 on Build GRCh38
rs972842675 Chr.19: 15353948-15353948 on Build GRCh38
rs945742648 Chr.19: 15354108-15354108 on Build GRCh38
rs375328807 Chr.19: 15354301-15354308 on Build GRCh38
rs984314233 Chr.19: 15354022-15354022 on Build GRCh38
rs745736952 Chr.19: 15354180-15354180 on Build GRCh38
rs1025718394 Chr.19: 15354236-15354236 on Build GRCh38
rs895125651 Chr.19: 15353924-15353930 on Build GRCh38
rs1038497233 Chr.19: 15354151-15354151 on Build GRCh38
rs200559663 Chr.19: 15353930-15353930 on Build GRCh38
rs953357183 Chr.19: 15353893-15353893 on Build GRCh38
rs530407902 Chr.19: 15354325-15354325 on Build GRCh38
rs965807672 Chr.19: 15353940-15353940 on Build GRCh38
rs573265198 Chr.19: 15354300-15354300 on Build GRCh38
rs1012772836 Chr.19: 15353926-15353927 on Build GRCh38
rs889002151 Chr.19: 15354153-15354153 on Build GRCh38
rs1028964537 Chr.19: 15353884-15353884 on Build GRCh38
rs1056871516 Chr.19: 15353916-15353917 on Build GRCh38
rs898589747 Chr.19: 15354196-15354197 on Build GRCh38
rs768349594 Chr.19: 15354388-15354388 on Build GRCh38
rs571168346 Chr.19: 15354035-15354035 on Build GRCh38
rs959881208 Chr.19: 15354395-15354395 on Build GRCh38
rs765396374 Chr.19: 15354339-15354339 on Build GRCh38
rs1008821242 Chr.19: 15354168-15354168 on Build GRCh38
rs967220207 Chr.19: 15354106-15354106 on Build GRCh38
rs766609095 Chr.19: 15354030-15354030 on Build GRCh38
rs930003144 Chr.19: 15354197-15354197 on Build GRCh38
rs796224929 Chr.19: 15353946-15353947 on Build GRCh38
rs948076385 Chr.19: 15354142-15354142 on Build GRCh38
rs796825140 Chr.19: 15353940-15353941 on Build GRCh38
rs574794490 Chr.19: 15353918-15353918 on Build GRCh38
rs963735646 Chr.19: 15354008-15354008 on Build GRCh38
rs1034525922 Chr.19: 15354285-15354285 on Build GRCh38
rs938578847 Chr.19: 15353865-15353865 on Build GRCh38
rs116690709 Chr.19: 15354346-15354346 on Build GRCh38
rs766451857 Chr.19: 15354296-15354296 on Build GRCh38
rs989883623 Chr.19: 15354079-15354079 on Build GRCh38
rs35397454 Chr.19: 15354366-15354367 on Build GRCh38
rs955813982 Chr.19: 15353963-15353963 on Build GRCh38
rs71782936 Chr.19: 15353929-15353930 on Build GRCh38
rs1035946764 Chr.19: 15354386-15354386 on Build GRCh38
rs1008923254 Chr.19: 15354297-15354297 on Build GRCh38
rs758221222 Chr.19: 15354258-15354258 on Build GRCh38

View Full Product Details