Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40232222-40232821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT2
Entrez Gene ID
208
Gene Name
AKT serine/threonine kinase 2
Gene Aliases
HIHGHH, PKBB, PKBBETA, PRKBB, RAC-BETA
UniGene
Hs.631535
Gene Chromosome Location
Chr.19: 40230317-40285531 on Build GRCh38
Transcripts
NM_001243027.2 NM_001243028.2 NM_001626.5 XM_011526614.1 XM_011526615.1 XM_011526616.1 XM_011526618.1 XM_011526619.1 XM_011526620.1 XM_017026470.1 XM_017026471.1

SNP Detail

SNP ID Chromosome Location
rs1048878008 Chr.19: 40232402-40232402 on Build GRCh38
rs959821983 Chr.19: 40232247-40232247 on Build GRCh38
rs149821698 Chr.19: 40232424-40232424 on Build GRCh38
rs906882635 Chr.19: 40232433-40232433 on Build GRCh38
rs915332931 Chr.19: 40232278-40232278 on Build GRCh38
rs766687564 Chr.19: 40232290-40232290 on Build GRCh38
rs746554574 Chr.19: 40232473-40232473 on Build GRCh38
rs148533978 Chr.19: 40232706-40232706 on Build GRCh38
rs575323266 Chr.19: 40232456-40232456 on Build GRCh38
rs1029462473 Chr.19: 40232699-40232699 on Build GRCh38
rs967843573 Chr.19: 40232735-40232735 on Build GRCh38
rs984690929 Chr.19: 40232785-40232785 on Build GRCh38
rs377623622 Chr.19: 40232607-40232607 on Build GRCh38
rs947787477 Chr.19: 40232609-40232609 on Build GRCh38
rs997532213 Chr.19: 40232690-40232690 on Build GRCh38
rs923638914 Chr.19: 40232593-40232593 on Build GRCh38
rs779462254 Chr.19: 40232415-40232415 on Build GRCh38
rs923607856 Chr.19: 40232386-40232386 on Build GRCh38
rs574929065 Chr.19: 40232401-40232401 on Build GRCh38
rs568980837 Chr.19: 40232582-40232582 on Build GRCh38
rs41301963 Chr.19: 40232512-40232512 on Build GRCh38
rs931909491 Chr.19: 40232560-40232560 on Build GRCh38
rs966152059 Chr.19: 40232762-40232762 on Build GRCh38
rs909104534 Chr.19: 40232789-40232789 on Build GRCh38
rs895423331 Chr.19: 40232696-40232696 on Build GRCh38
rs56327981 Chr.19: 40232781-40232781 on Build GRCh38
rs755489698 Chr.19: 40232397-40232397 on Build GRCh38
rs552261594 Chr.19: 40232611-40232611 on Build GRCh38
rs878904412 Chr.19: 40232754-40232754 on Build GRCh38
rs556763599 Chr.19: 40232436-40232436 on Build GRCh38
rs940855259 Chr.19: 40232795-40232795 on Build GRCh38
rs73555557 Chr.19: 40232538-40232538 on Build GRCh38
rs758179603 Chr.19: 40232425-40232425 on Build GRCh38
rs770689968 Chr.19: 40232531-40232531 on Build GRCh38
rs965545564 Chr.19: 40232463-40232463 on Build GRCh38
rs923917955 Chr.19: 40232318-40232318 on Build GRCh38
rs908931750 Chr.19: 40232778-40232778 on Build GRCh38
rs969188090 Chr.19: 40232775-40232775 on Build GRCh38
rs921941449 Chr.19: 40232776-40232776 on Build GRCh38
rs537029863 Chr.19: 40232249-40232249 on Build GRCh38
rs188407538 Chr.19: 40232364-40232364 on Build GRCh38
rs550780505 Chr.19: 40232572-40232572 on Build GRCh38
rs115411240 Chr.19: 40232598-40232598 on Build GRCh38
rs1142298 Chr.19: 40232698-40232698 on Build GRCh38
rs902067185 Chr.19: 40232681-40232682 on Build GRCh38
rs1011616040 Chr.19: 40232714-40232714 on Build GRCh38
rs953913715 Chr.19: 40232346-40232346 on Build GRCh38
rs117036685 Chr.19: 40232641-40232641 on Build GRCh38
rs999287491 Chr.19: 40232744-40232744 on Build GRCh38
rs946604261 Chr.19: 40232356-40232356 on Build GRCh38
rs1031505604 Chr.19: 40232759-40232759 on Build GRCh38
rs953550070 Chr.19: 40232520-40232520 on Build GRCh38
rs534945416 Chr.19: 40232349-40232349 on Build GRCh38
rs186585471 Chr.19: 40232559-40232559 on Build GRCh38
rs1030895516 Chr.19: 40232746-40232746 on Build GRCh38
rs1035691225 Chr.19: 40232457-40232457 on Build GRCh38
rs78257861 Chr.19: 40232658-40232658 on Build GRCh38
rs978113874 Chr.19: 40232312-40232312 on Build GRCh38
rs1049297201 Chr.19: 40232614-40232614 on Build GRCh38
rs909032864 Chr.19: 40232579-40232579 on Build GRCh38
rs555931423 Chr.19: 40232311-40232311 on Build GRCh38
rs1142297 Chr.19: 40232700-40232700 on Build GRCh38
rs145836031 Chr.19: 40232464-40232464 on Build GRCh38
rs976199560 Chr.19: 40232468-40232468 on Build GRCh38
rs900382192 Chr.19: 40232384-40232384 on Build GRCh38
rs1012462572 Chr.19: 40232501-40232501 on Build GRCh38
rs999658585 Chr.19: 40232751-40232751 on Build GRCh38
rs1004577585 Chr.19: 40232423-40232423 on Build GRCh38
rs868724745 Chr.19: 40232300-40232300 on Build GRCh38
rs976522593 Chr.19: 40232766-40232766 on Build GRCh38
rs771520219 Chr.19: 40232594-40232594 on Build GRCh38
rs564031685 Chr.19: 40232509-40232509 on Build GRCh38
rs931810053 Chr.19: 40232396-40232396 on Build GRCh38
rs570526739 Chr.19: 40232313-40232313 on Build GRCh38
rs1048911309 Chr.19: 40232568-40232568 on Build GRCh38
rs1022529138 Chr.19: 40232707-40232707 on Build GRCh38
rs1056204374 Chr.19: 40232411-40232411 on Build GRCh38
rs889242905 Chr.19: 40232710-40232710 on Build GRCh38
rs535649863 Chr.19: 40232753-40232753 on Build GRCh38
rs990972544 Chr.19: 40232267-40232267 on Build GRCh38
rs1029936884 Chr.19: 40232345-40232345 on Build GRCh38
rs1055463445 Chr.19: 40232672-40232672 on Build GRCh38
rs915163098 Chr.19: 40232355-40232355 on Build GRCh38
rs765587103 Chr.19: 40232417-40232419 on Build GRCh38
rs568766963 Chr.19: 40232729-40232729 on Build GRCh38
rs557497485 Chr.19: 40232757-40232757 on Build GRCh38
rs73042697 Chr.19: 40232521-40232521 on Build GRCh38
rs865820250 Chr.19: 40232380-40232380 on Build GRCh38
rs906919148 Chr.19: 40232655-40232655 on Build GRCh38
rs545870625 Chr.19: 40232791-40232791 on Build GRCh38
rs976070313 Chr.19: 40232556-40232556 on Build GRCh38
rs770057506 Chr.19: 40232715-40232718 on Build GRCh38
rs992194565 Chr.19: 40232605-40232605 on Build GRCh38

View Full Product Details