Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77209211-77209770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769436 c.454C>T p.Q152* Chr.9: 77209491-77209491 on Build GRCh38
4740407 c.401delA p.K135fs*9 Chr.9: 77209438-77209438 on Build GRCh38
6820216 c.439A>G p.I147V Chr.9: 77209476-77209476 on Build GRCh38
1463042 c.481C>T p.R161C Chr.9: 77209518-77209518 on Build GRCh38
6820215 c.439A>G p.I147V Chr.9: 77209476-77209476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557145575 Chr.9: 77209276-77209276 on Build GRCh38
rs550854692 Chr.9: 77209696-77209696 on Build GRCh38
rs979478868 Chr.9: 77209545-77209545 on Build GRCh38
rs935514183 Chr.9: 77209352-77209352 on Build GRCh38
rs745649244 Chr.9: 77209425-77209425 on Build GRCh38
rs866588207 Chr.9: 77209603-77209603 on Build GRCh38
rs775147536 Chr.9: 77209398-77209398 on Build GRCh38
rs994541792 Chr.9: 77209338-77209338 on Build GRCh38
rs191322820 Chr.9: 77209401-77209401 on Build GRCh38
rs932875242 Chr.9: 77209243-77209243 on Build GRCh38
rs776129728 Chr.9: 77209476-77209476 on Build GRCh38
rs780393987 Chr.9: 77209393-77209393 on Build GRCh38
rs768085521 Chr.9: 77209452-77209452 on Build GRCh38
rs190349790 Chr.9: 77209353-77209353 on Build GRCh38
rs761366962 Chr.9: 77209506-77209506 on Build GRCh38
rs750736185 Chr.9: 77209379-77209379 on Build GRCh38
rs375066283 Chr.9: 77209692-77209692 on Build GRCh38
rs912814136 Chr.9: 77209678-77209678 on Build GRCh38
rs940156003 Chr.9: 77209311-77209311 on Build GRCh38
rs202090510 Chr.9: 77209435-77209435 on Build GRCh38
rs888216364 Chr.9: 77209669-77209669 on Build GRCh38
rs930407905 Chr.9: 77209722-77209722 on Build GRCh38
rs765526339 Chr.9: 77209373-77209373 on Build GRCh38
rs777957165 Chr.9: 77209244-77209244 on Build GRCh38
rs752154880 Chr.9: 77209398-77209398 on Build GRCh38
rs755059038 Chr.9: 77209399-77209399 on Build GRCh38
rs1036112244 Chr.9: 77209325-77209325 on Build GRCh38
rs758860269 Chr.9: 77209387-77209387 on Build GRCh38
rs965620992 Chr.9: 77209313-77209313 on Build GRCh38
rs904834329 Chr.9: 77209390-77209390 on Build GRCh38
rs939354860 Chr.9: 77209535-77209535 on Build GRCh38
rs1037149374 Chr.9: 77209700-77209700 on Build GRCh38
rs1049187582 Chr.9: 77209341-77209341 on Build GRCh38
rs1057013343 Chr.9: 77209592-77209592 on Build GRCh38
rs1017925741 Chr.9: 77209299-77209299 on Build GRCh38
rs1021083498 Chr.9: 77209530-77209530 on Build GRCh38
rs536826378 Chr.9: 77209248-77209248 on Build GRCh38
rs775270304 Chr.9: 77209442-77209442 on Build GRCh38
rs921280658 Chr.9: 77209589-77209589 on Build GRCh38
rs1018033496 Chr.9: 77209674-77209674 on Build GRCh38
rs575104019 Chr.9: 77209578-77209578 on Build GRCh38
rs990034892 Chr.9: 77209357-77209357 on Build GRCh38
rs897572421 Chr.9: 77209337-77209337 on Build GRCh38
rs764790034 Chr.9: 77209507-77209507 on Build GRCh38
rs543626589 Chr.9: 77209687-77209687 on Build GRCh38
rs924181268 Chr.9: 77209340-77209340 on Build GRCh38
rs1007017718 Chr.9: 77209672-77209672 on Build GRCh38
rs772957048 Chr.9: 77209508-77209508 on Build GRCh38
rs919037125 Chr.9: 77209713-77209713 on Build GRCh38
rs1048775754 Chr.9: 77209744-77209744 on Build GRCh38
rs1034901281 Chr.9: 77209450-77209450 on Build GRCh38
rs757425143 Chr.9: 77209339-77209339 on Build GRCh38
rs777125490 Chr.9: 77209697-77209697 on Build GRCh38
rs372839456 Chr.9: 77209509-77209509 on Build GRCh38
rs541847519 Chr.9: 77209418-77209418 on Build GRCh38
rs755548637 Chr.9: 77209566-77209566 on Build GRCh38
rs762928129 Chr.9: 77209533-77209536 on Build GRCh38
rs897066324 Chr.9: 77209594-77209594 on Build GRCh38
rs987018735 Chr.9: 77209637-77209637 on Build GRCh38
rs766786397 Chr.9: 77209544-77209544 on Build GRCh38
rs915345397 Chr.9: 77209360-77209360 on Build GRCh38
rs576980714 Chr.9: 77209296-77209296 on Build GRCh38
rs371352398 Chr.9: 77209564-77209564 on Build GRCh38
rs1027211594 Chr.9: 77209633-77209633 on Build GRCh38
rs1009399787 Chr.9: 77209474-77209474 on Build GRCh38
rs954040603 Chr.9: 77209632-77209632 on Build GRCh38
rs370861429 Chr.9: 77209471-77209471 on Build GRCh38
rs185876418 Chr.9: 77209307-77209307 on Build GRCh38
rs376191694 Chr.9: 77209443-77209443 on Build GRCh38
rs770154538 Chr.9: 77209415-77209415 on Build GRCh38

View Full Product Details