Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127467687-127468284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
UniGene
Hs.495188
Gene Chromosome Location
Chr.9: 127451486-127503501 on Build GRCh38
Transcripts
NM_001005373.3 NM_001005374.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654425 c.619G>A p.E207K Chr.9: 127467830-127467830 on Build GRCh38
4552108 c.549G>A p.S183S Chr.9: 127467760-127467760 on Build GRCh38
3722435 c.588C>T p.G196G Chr.9: 127467799-127467799 on Build GRCh38
4699344 c.610C>A p.L204I Chr.9: 127467821-127467821 on Build GRCh38
118089 c.591T>G p.T197T Chr.9: 127467802-127467802 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542077582 Chr.9: 127467809-127467809 on Build GRCh38
rs376452859 Chr.9: 127468188-127468188 on Build GRCh38
rs983653252 Chr.9: 127468166-127468166 on Build GRCh38
rs770398029 Chr.9: 127467844-127467844 on Build GRCh38
rs956107977 Chr.9: 127467757-127467757 on Build GRCh38
rs564420811 Chr.9: 127467875-127467875 on Build GRCh38
rs138830549 Chr.9: 127467774-127467774 on Build GRCh38
rs199916476 Chr.9: 127467857-127467857 on Build GRCh38
rs780168494 Chr.9: 127467719-127467719 on Build GRCh38
rs375143572 Chr.9: 127467749-127467749 on Build GRCh38
rs373240847 Chr.9: 127468100-127468100 on Build GRCh38
rs767746171 Chr.9: 127467707-127467707 on Build GRCh38
rs974998243 Chr.9: 127467777-127467777 on Build GRCh38
rs760792213 Chr.9: 127467848-127467848 on Build GRCh38
rs755308288 Chr.9: 127467745-127467745 on Build GRCh38
rs533193448 Chr.9: 127467901-127467901 on Build GRCh38
rs759232309 Chr.9: 127467930-127467930 on Build GRCh38
rs756557431 Chr.9: 127467756-127467756 on Build GRCh38
rs947752458 Chr.9: 127467762-127467762 on Build GRCh38
rs755362254 Chr.9: 127467789-127467789 on Build GRCh38
rs761890031 Chr.9: 127467778-127467778 on Build GRCh38
rs148059394 Chr.9: 127467797-127467797 on Build GRCh38
rs1003384901 Chr.9: 127467723-127467723 on Build GRCh38
rs746274685 Chr.9: 127467815-127467815 on Build GRCh38
rs939064412 Chr.9: 127467969-127467969 on Build GRCh38
rs1030605138 Chr.9: 127468141-127468141 on Build GRCh38
rs754162095 Chr.9: 127467734-127467734 on Build GRCh38
rs919178344 Chr.9: 127468014-127468015 on Build GRCh38
rs770410548 Chr.9: 127467754-127467754 on Build GRCh38
rs562273106 Chr.9: 127467836-127467836 on Build GRCh38
rs371724522 Chr.9: 127467718-127467718 on Build GRCh38
rs980570870 Chr.9: 127467938-127467938 on Build GRCh38
rs370502826 Chr.9: 127467841-127467841 on Build GRCh38
rs182429667 Chr.9: 127468139-127468139 on Build GRCh38
rs148704769 Chr.9: 127467843-127467843 on Build GRCh38
rs774399646 Chr.9: 127467775-127467775 on Build GRCh38
rs754062373 Chr.9: 127467787-127467787 on Build GRCh38
rs1030792050 Chr.9: 127467939-127467939 on Build GRCh38
rs766431704 Chr.9: 127467860-127467860 on Build GRCh38
rs943176963 Chr.9: 127468099-127468099 on Build GRCh38
rs775020499 Chr.9: 127467764-127467764 on Build GRCh38
rs776877360 Chr.9: 127467864-127467864 on Build GRCh38
rs969563857 Chr.9: 127467934-127467934 on Build GRCh38
rs767583106 Chr.9: 127467779-127467779 on Build GRCh38
rs758777482 Chr.9: 127467877-127467877 on Build GRCh38
rs552266833 Chr.9: 127468016-127468016 on Build GRCh38
rs758867006 Chr.9: 127467804-127467804 on Build GRCh38
rs1018239202 Chr.9: 127467706-127467706 on Build GRCh38
rs2253669 Chr.9: 127468235-127468235 on Build GRCh38
rs779458921 Chr.9: 127467747-127467747 on Build GRCh38
rs888923362 Chr.9: 127468070-127468070 on Build GRCh38
rs745326217 Chr.9: 127467806-127467806 on Build GRCh38
rs964710578 Chr.9: 127468124-127468124 on Build GRCh38
rs75690855 Chr.9: 127467759-127467759 on Build GRCh38
rs780636250 Chr.9: 127467755-127467755 on Build GRCh38
rs762796023 Chr.9: 127467768-127467768 on Build GRCh38
rs142782210 Chr.9: 127467780-127467780 on Build GRCh38
rs144645671 Chr.9: 127467796-127467796 on Build GRCh38
rs972136840 Chr.9: 127468002-127468002 on Build GRCh38
rs201488957 Chr.9: 127467784-127467784 on Build GRCh38
rs368243544 Chr.9: 127467865-127467865 on Build GRCh38
rs141972828 Chr.9: 127468030-127468030 on Build GRCh38
rs955126594 Chr.9: 127468157-127468157 on Build GRCh38
rs765477712 Chr.9: 127467871-127467871 on Build GRCh38
rs975135121 Chr.9: 127468140-127468140 on Build GRCh38
rs141428822 Chr.9: 127467829-127467829 on Build GRCh38
rs927690600 Chr.9: 127467942-127467942 on Build GRCh38
rs999340012 Chr.9: 127467909-127467909 on Build GRCh38
rs545814024 Chr.9: 127467781-127467781 on Build GRCh38
rs892737252 Chr.9: 127468239-127468239 on Build GRCh38
rs144502205 Chr.9: 127468186-127468191 on Build GRCh38
rs886063455 Chr.9: 127467826-127467826 on Build GRCh38
rs191106111 Chr.9: 127468108-127468108 on Build GRCh38
rs773392393 Chr.9: 127467855-127467855 on Build GRCh38
rs1021596766 Chr.9: 127468050-127468050 on Build GRCh38
rs575482891 Chr.9: 127467940-127467940 on Build GRCh38

View Full Product Details