Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132725454-132725785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK8
Entrez Gene ID
158067
Gene Name
adenylate kinase 8
Gene Aliases
AK 8, C9orf98, DDX31
UniGene
Hs.421340
Gene Chromosome Location
Chr.9: 132725574-132878811 on Build GRCh38
Transcripts
NM_001317958.1 NM_001317959.1 NM_152572.2 XM_005272169.2 XM_006716965.2 XM_011518278.2 XM_017014308.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996407 c.1398C>G p.I466M Chr.9: 132725730-132725730 on Build GRCh38
7204585 c.1368C>G p.D456E Chr.9: 132725760-132725760 on Build GRCh38
4970070 c.1399G>A p.E467K Chr.9: 132725729-132725729 on Build GRCh38
4987079 c.1406G>A p.G469E Chr.9: 132725722-132725722 on Build GRCh38
5526469 c.1343G>A p.G448E Chr.9: 132725785-132725785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144348048 Chr.9: 132725709-132725709 on Build GRCh38
rs531715754 Chr.9: 132725536-132725536 on Build GRCh38
rs958370309 Chr.9: 132725521-132725521 on Build GRCh38
rs1048521054 Chr.9: 132725605-132725605 on Build GRCh38
rs1036169119 Chr.9: 132725754-132725754 on Build GRCh38
rs28477720 Chr.9: 132725739-132725739 on Build GRCh38
rs550749419 Chr.9: 132725573-132725573 on Build GRCh38
rs1052609389 Chr.9: 132725549-132725549 on Build GRCh38
rs770804748 Chr.9: 132725667-132725667 on Build GRCh38
rs988206339 Chr.9: 132725684-132725684 on Build GRCh38
rs199655656 Chr.9: 132725701-132725701 on Build GRCh38
rs764330683 Chr.9: 132725537-132725537 on Build GRCh38
rs767838160 Chr.9: 132725715-132725715 on Build GRCh38
rs979680875 Chr.9: 132725668-132725668 on Build GRCh38
rs979206707 Chr.9: 132725480-132725480 on Build GRCh38
rs189581069 Chr.9: 132725747-132725747 on Build GRCh38
rs745774866 Chr.9: 132725677-132725677 on Build GRCh38
rs931836603 Chr.9: 132725473-132725473 on Build GRCh38
rs373817929 Chr.9: 132725658-132725658 on Build GRCh38
rs1020103880 Chr.9: 132725630-132725630 on Build GRCh38
rs367966120 Chr.9: 132725745-132725745 on Build GRCh38
rs917622287 Chr.9: 132725552-132725552 on Build GRCh38
rs965374543 Chr.9: 132725631-132725631 on Build GRCh38
rs779703089 Chr.9: 132725746-132725747 on Build GRCh38
rs529986751 Chr.9: 132725637-132725637 on Build GRCh38
rs779977107 Chr.9: 132725639-132725639 on Build GRCh38
rs756005237 Chr.9: 132725745-132725746 on Build GRCh38
rs1032471093 Chr.9: 132725516-132725516 on Build GRCh38
rs375825588 Chr.9: 132725675-132725675 on Build GRCh38
rs138440349 Chr.9: 132725700-132725700 on Build GRCh38
rs976003191 Chr.9: 132725548-132725548 on Build GRCh38
rs372487313 Chr.9: 132725756-132725756 on Build GRCh38
rs1004334426 Chr.9: 132725627-132725627 on Build GRCh38
rs995686419 Chr.9: 132725574-132725574 on Build GRCh38
rs370791861 Chr.9: 132725685-132725685 on Build GRCh38
rs774005338 Chr.9: 132725705-132725705 on Build GRCh38
rs768062930 Chr.9: 132725704-132725704 on Build GRCh38
rs931896348 Chr.9: 132725554-132725554 on Build GRCh38
rs766978504 Chr.9: 132725729-132725729 on Build GRCh38
rs949018912 Chr.9: 132725569-132725569 on Build GRCh38
rs533440034 Chr.9: 132725742-132725742 on Build GRCh38
rs564533531 Chr.9: 132725487-132725487 on Build GRCh38
rs750837318 Chr.9: 132725718-132725718 on Build GRCh38
rs77647870 Chr.9: 132725641-132725641 on Build GRCh38
rs184476196 Chr.9: 132725730-132725730 on Build GRCh38
rs1050690053 Chr.9: 132725564-132725564 on Build GRCh38
rs779127805 Chr.9: 132725663-132725663 on Build GRCh38
rs1024618676 Chr.9: 132725628-132725628 on Build GRCh38
rs1045362769 Chr.9: 132725570-132725570 on Build GRCh38
rs306524 Chr.9: 132725625-132725625 on Build GRCh38
rs1028881427 Chr.9: 132725648-132725648 on Build GRCh38
rs376085711 Chr.9: 132725665-132725665 on Build GRCh38
rs200678908 Chr.9: 132725722-132725722 on Build GRCh38
rs148764112 Chr.9: 132725738-132725738 on Build GRCh38
rs368473946 Chr.9: 132725755-132725755 on Build GRCh38
rs754008893 Chr.9: 132725644-132725644 on Build GRCh38
rs887311396 Chr.9: 132725753-132725753 on Build GRCh38
rs781168820 Chr.9: 132725671-132725671 on Build GRCh38
rs944020786 Chr.9: 132725541-132725541 on Build GRCh38
rs897523493 Chr.9: 132725620-132725620 on Build GRCh38
rs769479139 Chr.9: 132725751-132725751 on Build GRCh38

View Full Product Details