Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50192512-50193098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP4
Entrez Gene ID
8852
Gene Name
A-kinase anchoring protein 4
Gene Aliases
AKAP 82, AKAP-4, AKAP82, CT99, FSC1, HI, PRKA4, hAKAP82, p82
UniGene
Hs.97633
Gene Chromosome Location
Chr.X: 50190768-50201013 on Build GRCh38
Transcripts
NM_003886.2 NM_139289.1

SNP Detail

SNP ID Chromosome Location
rs139049075 Chr.X: 50193055-50193055 on Build GRCh38
rs201649248 Chr.X: 50193038-50193038 on Build GRCh38
rs782299034 Chr.X: 50193030-50193030 on Build GRCh38
rs782124961 Chr.X: 50192879-50192879 on Build GRCh38
rs782618799 Chr.X: 50192714-50192714 on Build GRCh38
rs782297269 Chr.X: 50192710-50192710 on Build GRCh38
rs782676793 Chr.X: 50192703-50192703 on Build GRCh38
rs781839360 Chr.X: 50192950-50192950 on Build GRCh38
rs12012704 Chr.X: 50192695-50192695 on Build GRCh38
rs150728490 Chr.X: 50192890-50192890 on Build GRCh38
rs782248153 Chr.X: 50192998-50192998 on Build GRCh38
rs781948063 Chr.X: 50192815-50192815 on Build GRCh38
rs782474652 Chr.X: 50192952-50192952 on Build GRCh38
rs782392195 Chr.X: 50192723-50192723 on Build GRCh38
rs782658466 Chr.X: 50192694-50192694 on Build GRCh38
rs143933731 Chr.X: 50193064-50193064 on Build GRCh38
rs782296919 Chr.X: 50192555-50192555 on Build GRCh38
rs782159385 Chr.X: 50192754-50192754 on Build GRCh38
rs930232562 Chr.X: 50193071-50193071 on Build GRCh38
rs782020421 Chr.X: 50192709-50192709 on Build GRCh38
rs782311961 Chr.X: 50192780-50192780 on Build GRCh38
rs782649504 Chr.X: 50192964-50192964 on Build GRCh38
rs142687955 Chr.X: 50192872-50192872 on Build GRCh38
rs782215128 Chr.X: 50192716-50192716 on Build GRCh38
rs138583730 Chr.X: 50192825-50192825 on Build GRCh38
rs967320435 Chr.X: 50192730-50192730 on Build GRCh38
rs138199562 Chr.X: 50192557-50192557 on Build GRCh38
rs782708684 Chr.X: 50192677-50192677 on Build GRCh38
rs782053667 Chr.X: 50192568-50192568 on Build GRCh38
rs182407156 Chr.X: 50192691-50192691 on Build GRCh38
rs781961150 Chr.X: 50193043-50193043 on Build GRCh38
rs782186826 Chr.X: 50193028-50193028 on Build GRCh38
rs782672648 Chr.X: 50193019-50193019 on Build GRCh38
rs373820674 Chr.X: 50193017-50193017 on Build GRCh38
rs200939746 Chr.X: 50192688-50192688 on Build GRCh38
rs191821579 Chr.X: 50192663-50192663 on Build GRCh38
rs782127705 Chr.X: 50192638-50192638 on Build GRCh38
rs1132161 Chr.X: 50193034-50193034 on Build GRCh38
rs782820292 Chr.X: 50192908-50192908 on Build GRCh38
rs147525170 Chr.X: 50192768-50192768 on Build GRCh38
rs782467443 Chr.X: 50192920-50192920 on Build GRCh38
rs782551339 Chr.X: 50193005-50193005 on Build GRCh38
rs370374358 Chr.X: 50192921-50192921 on Build GRCh38
rs781979488 Chr.X: 50192749-50192749 on Build GRCh38
rs12011833 Chr.X: 50192565-50192565 on Build GRCh38
rs782630376 Chr.X: 50192552-50192552 on Build GRCh38
rs374992303 Chr.X: 50192614-50192614 on Build GRCh38
rs143213342 Chr.X: 50192722-50192722 on Build GRCh38
rs1047259655 Chr.X: 50192582-50192582 on Build GRCh38
rs782396822 Chr.X: 50192766-50192766 on Build GRCh38
rs781891685 Chr.X: 50192913-50192913 on Build GRCh38
rs782773415 Chr.X: 50192648-50192648 on Build GRCh38
rs143297000 Chr.X: 50192674-50192674 on Build GRCh38

View Full Product Details